Product References
Differential expression of the insulin-like growth factor receptor among early breast cancer subtypes.
PloS one
Mountzios G,Aivazi D,Kostopoulos I,Kourea HP,Kouvatseas G,Timotheadou E,Zebekakis P,Efstratiou I,Gogas H,Vamvouka C,Chrisafi S,Stofas A,Pentheroudakis G,Koutras A,Galani E,Bafaloukos D,Fountzilas G
MA5-13802 was used in immunohistochemistry to measure protein expression of insulin-like growth factor receptor pathway members and evaluate their prognostic significance across the different early breast cancer subtypes
Tue Jan 06 00:00:00 EST 2015
Immunohistochemical analysis of RTKs expression identified HER3 as a prognostic indicator of gastric cancer.
Cancer science
Ema A,Yamashita K,Ushiku H,Kojo K,Minatani N,Kikuchi M,Mieno H,Moriya H,Hosoda K,Katada N,Kikuchi S,Watanabe M
MA5-13802 was used in immunohistochemistry - paraffin section to determine if receptor type tyrosine kinases have prognostic relevance in patients with stage II/III gastric cancer treated with standard treatment
Mon Dec 01 00:00:00 EST 2014
Insulin-like growth factor 1 receptor (IGF1R) expression and survival in operable squamous-cell laryngeal cancer.
PloS one
Mountzios G,Kostopoulos I,Kotoula V,Sfakianaki I,Fountzilas E,Markou K,Karasmanis I,Leva S,Angouridakis N,Vlachtsis K,Nikolaou A,Konstantinidis I,Fountzilas G
MA5-13802 was used in immunohistochemistry to study the prognostic value of IGF-1R expression in operable squamous cell laryngeal cancer
Mon Jul 08 00:00:00 EDT 2013
Utility of insulin-like growth factor receptor-1 expression in gefitinib-treated patients with non-small cell lung cancer.
Anticancer research
Fidler MJ,Basu S,Buckingham L,Walters K,McCormack S,Batus M,Coon J,Bonomi P
MA5-13802 was used in immunohistochemistry to study whether IGF1R expression in non-small cell lung cancer patients is linked to gefitinib resistance
Tue May 01 00:00:00 EDT 2012
Does insulin-like growth factor 1 receptor (IGF-1R) targeting provide new treatment options for chordomas? A retrospective clinical and immunohistochemical study.
Histopathology
Scheipl S,Froehlich EV,Leithner A,Beham A,Quehenberger F,Mokry M,Stammberger H,Varga PP,Lazáry A,Windhager R,Gattenloehner S,Liegl B
MA5-13802 was used in immunohistochemistry to study the therapeutic potential of targeting IGF-1R in chordoma
Tue May 01 00:00:00 EDT 2012
Phase II study of preoperative systemic treatment with the combination of docetaxel and trastuzumab in patients with locally advanced HER-2-overexpressing breast cancer.
Breast (Edinburgh, Scotland)
Sawaki M,Iwata H,Sato Y,Wada M,Toyama T,Sasaki E,Yatabe Y,Imai T,Ohashi Y
MA5-13802 was used in immunohistochemistry to investigate the efficacy of docetaxel and trastuzumab against locally advanced HER-2-overexpressing breast cancer
Fri Oct 01 00:00:00 EDT 2010
High coexpression of both insulin-like growth factor receptor-1 (IGFR-1) and epidermal growth factor receptor (EGFR) is associated with shorter disease-free survival in resected non-small-cell lung cancer patients.
Annals of oncology : official journal of the European Society for Medical Oncology
Ludovini V,Bellezza G,Pistola L,Bianconi F,Di Carlo L,Sidoni A,Semeraro A,Del Sordo R,Tofanetti FR,Mameli MG,Daddi G,Cavaliere A,Tonato M,Crinò L
MA5-13802 was used in immunohistochemistry to investigate the prognostic significance of IGF-1 receptor and EGF receptor expression in resected non-small-cell lung cancer patients
Fri May 01 00:00:00 EDT 2009
Relationships of insulin-like growth factor-1 receptor and epidermal growth factor receptor expression to clinical outcomes in patients with colorectal cancer.
Oncology
Takahari D,Yamada Y,Okita NT,Honda T,Hirashima Y,Matsubara J,Takashima A,Kato K,Hamaguchi T,Shirao K,Shimada Y,Shimoda T
MA5-13802 was used in immunohistochemistry to investigate the prognostic significance of IGF-1 receptor and EGF receptor expression in colorectal cancer
Mon Jan 26 00:00:00 EST 2009
Impact of insulin-like growth factor type 1 receptor, epidermal growth factor receptor, and HER2 expressions on outcomes of patients with gastric cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research
Matsubara J,Yamada Y,Hirashima Y,Takahari D,Okita NT,Kato K,Hamaguchi T,Shirao K,Shimada Y,Shimoda T
MA5-13802 was used in immunohistochemistry to study the prognostic significance of IGF-1 receptor, HER2, and EGF receptor in gastric cancer
Thu May 15 00:00:00 EDT 2008
Intratumoral IGF-I protein expression is selectively upregulated in breast cancer patients with BRCA1/2 mutations.
Endocrine-related cancer
Hudelist G,Wagner T,Rosner M,Fink-Retter A,Gschwantler-Kaulich D,Czerwenka K,Kroiss R,Tea M,Pischinger K,Köstler WJ,Attems J,Mueller R,Blaukopf C,Kubista E,Hengstschläger M,Singer CF
MA5-13802 was used in immunohistochemistry to investigate the expression of intratumoral IGF-I protein in breast cancer patients with BRCA1/2 mutations
Sat Dec 01 00:00:00 EST 2007
Semi-quantitative scoring of potentially predictive markers for endocrine treatment of breast cancer: a comparison between whole sections and tissue microarrays.
Journal of clinical pathology
Henriksen KL,Rasmussen BB,Lykkesfeldt AE,Møller S,Ejlertsen B,Mouridsen HT
MA5-13802 was used in immunohistochemistry to study the use of tissue microarrays for the semi-quantitative scoring of potentially predictive markers for endocrine treatment of breast cancer
Sun Apr 01 00:00:00 EDT 2007
Predictors of resistance to preoperative trastuzumab and vinorelbine for HER2-positive early breast cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research
Harris LN,You F,Schnitt SJ,Witkiewicz A,Lu X,Sgroi D,Ryan PD,Come SE,Burstein HJ,Lesnikoski BA,Kamma M,Friedman PN,Gelman R,Iglehart JD,Winer EP
MA5-13802 was used in immunohistochemistry to study predictors of response to pre-operative trastuzumab and vinorelbine for HER2-positive early breast cancer
Thu Feb 15 00:00:00 EST 2007
Multiple bronchioloalveolar carcinomas in acromegaly: a potential role of insulin-like growth factor I in carcinogenesis.
Lung cancer (Amsterdam, Netherlands)
Motoi N,Kishi K,Fujii T,Tsuboi E,Ohashi K,Yoshimura K
MA5-13802 was used in immunohistochemistry to investigate the novel role of insulin-like growth factor I in carcinogenesis
Wed Nov 01 00:00:00 EST 2006
The p53 pathway and outcome among patients with T1G3 bladder tumors.
Clinical cancer research : an official journal of the American Association for Cancer Research
López-Knowles E,Hernández S,Kogevinas M,Lloreta J,Amorós A,Tardón A,Carrato A,Kishore S,Serra C,Malats N,Real FX
MA5-13802 was used in immunohistochemistry to study the prognostic value of p53 pathway expression in patients with T1G3 bladder tumors
Sun Oct 15 00:00:00 EDT 2006
Increased hepatic expression of insulin-like growth factor-I receptor in chronic hepatitis C.
World journal of gastroenterology
Stefano JT,Correa-Giannella ML,Ribeiro CM,Alves VA,Massarollo PC,Machado MC,Giannella-Neto D
MA5-13802 was used in immunohistochemistry to investigate expression of insulin-like growth factor-I receptor and insulin-like growth factor-I in livers from patients with chronic hepatitis C
Wed Jun 28 00:00:00 EDT 2006
Insulin-like growth factor-1 receptor (IGF-1R) expression does not predict for resistance to trastuzumab-based treatment in patients with Her-2/neu overexpressing metastatic breast cancer.
Journal of cancer research and clinical oncology
Köstler WJ,Hudelist G,Rabitsch W,Czerwenka K,Müller R,Singer CF,Zielinski CC
MA5-13802 was used in immunohistochemistry to evaluate the clinical relevance of insulin-like growth factor-1 receptor expression within the context of trastuzumab-based therapy
Sun Jan 01 00:00:00 EST 2006
Retinoblastoma susceptibility gene product pRB activates hypoxia-inducible factor-1 (HIF-1).
Oncogene
Budde A,Schneiderhan-Marra N,Petersen G,Brüne B
MA5-13802 was used in ChIP assay to study the role of retinoblastoma susceptibility gene product I in hypoxia-inducible factor-1 induction
Thu Mar 03 00:00:00 EST 2005
The pT1a and pT1b category subdivision in renal cell carcinoma: is it reflected by differences in tumour biology?
BJU international
Langner C,Ratschek M,Rehak P,Tsybrovskyy O,Zigeuner R
MA5-13802 was used in immunohistochemistry to study the category subdivision of pT1a and pT1b in renal cell carcinoma
Tue Feb 01 00:00:00 EST 2005
Expression of tyrosine kinases in human malignancies as potential targets for kinase-specific inhibitors.
Endocrine-related cancer
Singer CF,Hudelist G,Lamm W,Mueller R,Czerwenka K,Kubista E
MA5-13802 was used in immunohistochemistry to study the expression of tyrosine kinases in human malignancies as potential targets for kinase-specific inhibitors
Wed Dec 01 00:00:00 EST 2004
Analysis of insulin-like growth factors and insulin-like growth factor I receptor expression in renal cell carcinoma.
American journal of clinical pathology
Schips L,Zigeuner R,Ratschek M,Rehak P,Rüschoff J,Langner C
MA5-13802 was used in immunohistochemistry to study the expression of IGF-IR and IGF I and II in renal cell carcinoma
Wed Dec 01 00:00:00 EST 2004
The role of beta-catenin, TGF beta 3, NGF2, FGF2, IGFR2, and BMP4 in the pathogenesis of mesenteric sclerosis and angiopathy in midgut carcinoids.
Human pathology
Zhang PJ,Furth EE,Cai X,Goldblum JR,Pasha TL,Min KW
MA5-13802 was used in immunohistochemistry to study the role of several fibrosing-related growth factor in the pathogenesis of mesenteric sclerosis and angiopathy in midgut carcinoids
Tue Jun 01 00:00:00 EDT 2004
Expression of insulin-like growth factor-I receptor in primary cutaneous carcinomas.
Journal of cutaneous pathology
Keehn CA,Saeed S,Bickle K,Khalil FK,Morgan MB
MA5-13802 was used in immunohistochemistry to study the role of insulin-like growth factor-I receptor in the carcinogenesis of primary cutaneous carcinomas
Sat May 01 00:00:00 EDT 2004
Coexpression of IGF-1R and c-Src proteins in human pancreatic ductal adenocarcinoma.
Digestive diseases and sciences
Hakam A,Fang Q,Karl R,Coppola D
MA5-13802 was used in immunohistochemistry to show the co-localization of IGF-1R and c-Src in human pancreatic ductal adenocarcinoma
Wed Oct 01 00:00:00 EDT 2003