Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • IHH Antibodies

          Proteintech

          IHH Polyclonal Antibody

          9 References
          View all (15) IHH antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite IHH Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (8)
          IHH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          IHH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IHH Antibody (13388-1-AP) in IHC (P)

          Immunohistochemistry of paraffin-embedded human stomach cancer tissue slide using 13388-1-AP (IHH Antibody) at dilution of 1:200 (under 40x lens). heat mediated antigen retrieved with Tris-EDTA buffer (pH 9). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IHH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IHH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IHH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IHH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IHH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IHH Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          IHH Antibody in Western Blot (WB)
          IHH Antibody in Western Blot (WB)
          IHH Polyclonal Antibody

          Product Details

          13388-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 5 publications 5 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:400
          -

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          IHH Fusion Protein Ag4046 (103-411 aa encoded by BC034757)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.24 mg/mL

          Amount

          36 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: TQRCKDRLN SLAISVMNQW PGVKLRVTEG WDEDGHHSEE SLHYEGRAVD ITTSDRDRNK YGLLARLAVE AGFDWVYYES KAHVHCSVKS EHSAAAKTGG CFPAGAQVRL ESGARVALSA VRPGDRVLAM GEDGSPTFSD VLIFLDREPH RLRAFQVIET QDPPRRLALT PAHLLFTADN HTEPAARFRA TFASHVQPGQ YVLVAGVPGL QPARVAAVST HVALGAYAPL TKHGTLVVED VVASCFAAVA DHHLAQLAFW PLRLFHSLAW GSWTPGEGVH WYPQLLYRLG RLLLEEGSFH PLGMSGAGS (103-411 aa encoded by BC034757)

          Target Information

          Ihh encodes a member of the hedgehog family of secreted signaling molecules. Hedgehog proteins are essential regulators of a variety of developmental processes including growth, patterning and morphogenesis. The encoded protein specifically plays a role in bone growth an differentiation. Mutations in this gene are the cause of brachydactyly type A1 which is characterized by shortening or malformation of the phalanges. Mutations in this gene are also the cause of acrocapitofemoral dysplasia.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: HHG-2; IHH; Indian hedgehog homolog; Indian hedgehog protein

          View more View less

          Gene Aliases: BDA1; HHG2; IHH

          View more View less

          UniProt ID: (Human) Q14623, (Mouse) P97812

          View more View less

          Entrez Gene ID: (Human) 3549, (Mouse) 16147, (Rat) 84399

          View more View less

          Function(s)
          patched binding calcium ion binding protein binding peptidase activity transferase activity hydrolase activity very-low-density lipoprotein particle binding metal ion binding cholesterol-protein transferase activity molecular_function
          Process(es)
          skeletal system development ossification patterning of blood vessels osteoblast differentiation in utero embryonic development cell fate specification morphogenesis of a branching structure vasculature development heart looping positive regulation of mesenchymal cell proliferation epithelial cell morphogenesis retinal pigment epithelium development proteoglycan metabolic process proteolysis cell communication smoothened signaling pathway cell-cell signaling multicellular organism development pattern specification process positive regulation of cell proliferation embryo development embryonic pattern specification negative regulation of signal transduction intein-mediated protein splicing cell differentiation vitelline membrane formation pancreas development positive regulation of chondrocyte differentiation positive regulation of collagen biosynthetic process negative regulation of T cell differentiation in thymus negative regulation of immature T cell proliferation in thymus positive regulation of T cell differentiation in thymus multicellular organism growth chondrocyte proliferation regulation of growth embryonic digit morphogenesis camera-type eye development negative regulation of apoptotic process bone resorption regulation of cell differentiation negative regulation of cell differentiation positive regulation of smoothened signaling pathway positive regulation of transcription from RNA polymerase II promoter positive regulation of alpha-beta T cell differentiation negative regulation of alpha-beta T cell differentiation negative regulation of eye pigmentation cell maturation embryonic digestive tract morphogenesis embryonic camera-type eye morphogenesis neuron development smooth muscle tissue development positive regulation of epithelial cell proliferation cartilage development camera-type eye photoreceptor cell fate commitment head morphogenesis somite development embryonic skeletal joint development epithelial cell-cell adhesion response to estradiol maternal process involved in female pregnancy chondrocyte differentiation chondrocyte differentiation involved in endochondral bone morphogenesis cell proliferation response to mechanical stimulus regulation of gene expression protein autoprocessing negative regulation of chondrocyte differentiation system development self proteolysis liver regeneration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline