Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • IL-10 Antibodies

          Invitrogen

          IL-10 Polyclonal Antibody, PeproTech®

          2 Published Figures
          12 References
          View all (93) IL-10 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite IL-10 Polyclonal Antibody, PeproTech®

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (2)
          IL-10 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          IL-10 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IL-10 Antibody (500-P20-100UG) in WB

          Western Blot: To detect Human IL-10 by Western Blot analysis IL-10 Polyclonal Antibody (Product # 500-P20-1MG) can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human IL-10 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IL-10 Antibody in Western Blot (WB)
          IL-10 Antibody in Western Blot (WB)
          IL-10 Antibody in ELISA (ELISA)
          IL-10 Antibody in Neutralization (Neu)
          IL-10 Antibody in Neutralization (Neu)
          IL-10 Polyclonal Antibody, PeproTech®

          Product Details

          500-P20-100UG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.2 µg/mL
          -

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          0.5-2.0 µg/mL
          View 3 publications 3 publications

          Neutralization (Neu)

          -
          View 3 publications 3 publications

          In vitro Assay (In vitro)

          -
          View 3 publications 3 publications

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication

          Immunostaining (IS)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human

          Host/Isotype

          Rabbit

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E.coli-derived Recombinant Human IL-10
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.1-1.0 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          RRID

          AB_2929789

          Product Specific Information

          AA Sequence of recombinant protein: MSPGQGTQSE NSCTHFPGNL PNMLRDLRDA FSRVKTFFQM KDQLDNLLLK ESLLEDFKGY LGCQALSEMI QFYLEEVMPQ AENQDPDIKA HVNSLGENLK TLRLRLRRCH RFLPCENKSK AVEQVKNAFN KLQEKGIYKA MSEFDIFINY IEAYMTMKIR N.

          Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-10. Anti-Human IL-10-specific antibody was purified by affinity chromatography employing an immobilized Human IL-10 matrix.

          Sandwich ELISA: To detect Human IL-10 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.5-2.0 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human IL-10 (500-P20Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IL-10.

          Western Blot: To detect Human IL-10 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human IL-10 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

          Target Information

          IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CSIF; Cytokine synthesis inhibitory factor; H-IL-10; il 10; IL-10; IL10; ILN; Interleukin; Interleukin-10; Interleukin10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor

          View more View less

          Gene Aliases: CSIF; GVHDS; IL-10; IL10; IL10A; TGIF

          View more View less

          UniProt ID: (Human) P22301

          View more View less

          Entrez Gene ID: (Human) 3586

          View more View less

          Function(s)
          cytokine activity interleukin-10 receptor binding protein binding growth factor activity protein dimerization activity
          Process(es)
          negative regulation of cytokine production positive regulation of cytokine production positive regulation of endothelial cell proliferation response to molecule of bacterial origin chronic inflammatory response to antigenic stimulus positive regulation of immunoglobulin production negative regulation of cytokine production involved in immune response negative regulation of chronic inflammatory response to antigenic stimulus positive regulation of B cell apoptotic process immune response negative regulation of cell proliferation response to xenobiotic stimulus regulation of gene expression negative regulation of autophagy response to activity response to inactivity hemopoiesis B cell differentiation leukocyte chemotaxis negative regulation of myeloid dendritic cell activation negative regulation of B cell proliferation response to lipopolysaccharide negative regulation of interferon-alpha production negative regulation of interferon-gamma production negative regulation of interleukin-1 production negative regulation of interleukin-12 production negative regulation of interleukin-18 production negative regulation of interleukin-6 production negative regulation of interleukin-8 production negative regulation of tumor necrosis factor production response to insulin negative regulation of heterotypic cell-cell adhesion positive regulation of heterotypic cell-cell adhesion response to carbon monoxide cellular response to hepatocyte growth factor stimulus type 2 immune response B cell proliferation negative regulation of T cell proliferation defense response to bacterium defense response to protozoan positive regulation of macrophage activation negative regulation of apoptotic process negative regulation of neuron apoptotic process negative regulation of nitric oxide biosynthetic process regulation of isotype switching negative regulation of MHC class II biosynthetic process positive regulation of MHC class II biosynthetic process positive regulation of cell cycle positive regulation of transcription, DNA-templated negative regulation of mitotic cell cycle positive regulation of transcription from RNA polymerase II promoter positive regulation of JAK-STAT cascade negative regulation of inflammatory response regulation of synapse organization negative regulation of membrane protein ectodomain proteolysis response to glucocorticoid negative regulation of cytokine activity branching involved in labyrinthine layer morphogenesis cellular response to lipopolysaccharide cellular response to estradiol stimulus negative regulation of chemokine (C-C motif) ligand 5 production endothelial cell apoptotic process liver regeneration interleukin-10-mediated signaling pathway positive regulation of plasma cell differentiation positive regulation of pri-miRNA transcription from RNA polymerase II promoter regulation of response to wounding negative regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway regulation of complement-dependent cytotoxicity positive regulation of sprouting angiogenesis negative regulation of vascular smooth muscle cell proliferation positive regulation of vascular smooth muscle cell proliferation negative regulation of endothelial cell apoptotic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline