Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Description: This 8CH monoclonal antibody reacts with human IL-8 (CXCL8), a pro-inflammatory CXC chemokine. It is synthesized as a 99 amino acid precursor protein that is further processed into one of four isoforms, with the most common being 72 or 77 amino acids in length. IL-8(77) is secreted primarily by endothelial cells and is thought to be a less potent neutrophil activator than the other forms. It is present at high levels during fetal development, where it mediates angiogenesis rather than inflammation. The predominant form present in adults is IL-8(72), which is expressed by monocytes, macrophages, epithelial cells, and fibroblasts in response to inflammatory stimuli, environmental stress, and steroid hormones. IL-8(72) is essential for the activation and recruitment of neutrophils to sites of inflammation, and has also been found to influence T cell migration. Signaling occurs through the G-protein coupled receptors CXCR1 or CXCR2. IL-8 transcripts are often upregulated in tumors, and it is associated with tumor angiogenesis and metastasis.
Applications Reported: This 8CH antibody has been reported for use in intracellular staining followed by flow cytometric analysis.
Applications Tested: This 8CH antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis. This can be used at 5 µL (0.015 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.
Filtration: 0.2 µm post-manufacturing filtered.
Interleukin 8 (IL-8, CXCL8) is a 72 amino acid pro-inflammatory factor which belongs to the CXC subfamily of chemokines, and are bound by the cell surface receptors IL-8RA and IL-8RB. IL-8 functions as a chemoattractant and potent angiogenic factor. The expression and secretion of IL-8 can be induced by diverse inflammatory stimuli in many cells, including macrophages and endothelial cells. In endothelial cells, IL-8 is present in storage vesicles called Weibel-Palade bodies. IL-8, first isolated from osteosarcoma cells, contains the ELR-motif (N-terminal Glu-Leu-Arg amino acid sequence) and signals through the CXCR1 and CXCR2 receptors. Previous nomenclature for IL-8 includes neutrophil activating protein 1 (NAP-1), granulocyte chemotactic protein 1 (GCP-1), monocyte-derived neutrophil-activating peptide (MONAP) and protein 3-10C. IL-8 and ten other members of the CXC chemokine gene family form a chemokine gene cluster in a region mapped to chromosome 4q. Cancer studies have demonstrated a role for IL-8 in the angiogenesis and growth of tumours, and IL-8 is believed to play a role in the pathogenesis of bronchiolitis, a common respiratory tract disease caused by viral infection.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support