Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MVLLWLTLLL IALPCLLQTK EDPNPPITNL RMKAKAQQLT WDLNRNVTDI ECVKDADYSM PAVNNSYCQF GAISLCEVTN YTVRVANPPF STWILFPENS GKPWAGAENL TCWIHDVDFL SCSWAVGPGA PADVQYDLYL NVANRRQQYE CLHYKTDAQG TRIGCRFDDI SRLSSGSQSS HILVRGRSAA FGIPCTDKFV VFSQIEILTP PNMTAKCNKT HSFMHWKMRS HFNRKFRYEL QIQKRMQPVI TEQVRDRTSF QLLNPGTYTV QIRARERVYE FLSAWSTPQR FECDQEEGAN TRAWRTSLLI ALGTLLALVC VFVICRRYLV MQRLFPRIPH MKDPIGDSFQ NDKLVVWEAG KAGLEECLVT EVQVVQKT
IL-3Ra (Interleukin-3 receptor subunit alpha) belongs to the type I cytokine receptor family and is a heterodimer with a unique alpha chain paired with the common beta (beta c or CDw131) subunit. IL-3Ra is responsible for the transmission of signal of Interleukin-3. IL-3Ra and IL-3Rb chain heterodimerization is induced by Interleukin-3, which is required for receptor activation but not high affinity binding. IL-3Ra is also reported that IL-3Ra subunits are overexpressed on acute myeloid leukemia (AML) blasts compared with normal hematopoietic cells and are thus potential targets for novel therapeutic agents.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD123; CD123 antigen; IL-3 receptor alpha SP2 isoform; IL-3 receptor subunit alpha; IL-3R subunit alpha; IL-3R-alpha; IL-3RA; interleukin 3 receptor, alpha (low affinity); Interleukin-3 receptor subunit alpha
Gene Aliases: CD123; hIL-3Ra; IL3R; IL3RA; IL3RAY; IL3RX; IL3RY
UniProt ID: (Human) P26951
Entrez Gene ID: (Human) 3563
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support