Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • LILRB4 (CD85k) Antibodies

          Proteintech

          ILT-3/LILRB4 Polyclonal Antibody

          View all (19) LILRB4 (CD85k) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite ILT-3/LILRB4 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          ILT-3/LILRB4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ILT-3/LILRB4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ILT-3/LILRB4 Antibody (28422-1-AP) in WB

          Mouse heart tissue were subjected to SDS PAGE followed by western blot with 28422-1-AP (LILRB4 antibody) at dilution of 1:1,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ILT-3/LILRB4 Antibody in Western Blot (WB)
          ILT-3/LILRB4 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          ILT-3/LILRB4 Polyclonal Antibody

          Product Details

          28422-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:4,000
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ILT-3/LILRB4 fusion protein Ag28928
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.8 mg/mL

          Amount

          120 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: TLEAREYRLD KEESPAPWDR QNPLEPKNKA RFSIPSMTED YAGRYRCYYR SPVGWSQPSD PLELVMTGAY SKPTLSALPS PLVTSGKSVT LLCQSRSPMD TFLLIKERAA HPLLHLRSEH GAQQH

          Target Information

          This gene is a member of the leukocyte immunoglobulin-like receptor (LIR) family, which is found in a gene cluster at chromosomal region 19q13.4. The encoded protein belongs to the subfamily B class of LIR receptors which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and two to four cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The receptor is expressed on immune cells where it binds to MHC class I molecules on antigen-presenting cells and transduces a negative signal that inhibits stimulation of an immune response. The receptor can also function in antigen capture and presentation. It is thought to control inflammatory responses and cytotoxicity to help focus the immune response and limit autoreactivity. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2008].

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CD85 antigen-like family member K; glycoprotein 49 B; immunoglobulin-like transcript 3; leucocyte Ig-like receptor B4; leukocyte immunoglobulin-like receptor 5; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 4; leukocyte immunoglobulin-like receptor, subfamily B, member 4; LILRB4; mast cell surface glycoprotein Gp49B; monocyte inhibitory receptor HM18; unnamed protein product

          View more View less

          Gene Aliases: B4; CD85K; gp49; Gp49b; HM18; ILT-3; ILT3; Lilrb4; LIR-5; LIR5

          View more View less

          UniProt ID: (Human) Q8NHJ6, (Mouse) Q64281

          View more View less

          Entrez Gene ID: (Human) 11006, (Mouse) 14728

          View more View less

          Function(s)
          fibronectin binding protein binding protein phosphatase binding protein tyrosine kinase inhibitor activity transmembrane receptor protein tyrosine kinase inhibitor activity receptor inhibitor activity inhibitory MHC class I receptor activity apolipoprotein binding transmembrane signaling receptor activity integrin binding phosphatase binding immune receptor activity
          Process(es)
          neutrophil homeostasis inflammatory response to antigenic stimulus negative regulation of immunoglobulin production immune response-inhibiting cell surface receptor signaling pathway inflammatory response blood coagulation response to wounding response to nematode response to lipopolysaccharide negative regulation of natural killer cell activation negative regulation of mast cell activation negative regulation of mast cell activation involved in immune response CD4-positive, alpha-beta T cell proliferation CD8-positive, alpha-beta T cell proliferation type 2 immune response response to other organism cellular response to lipopolysaccharide negative regulation of monocyte activation negative regulation of plasma cell differentiation tolerance induction positive regulation of T cell anergy negative regulation of T cell cytokine production Fc receptor mediated inhibitory signaling pathway cell surface receptor signaling pathway cytokine-mediated signaling pathway receptor internalization negative regulation of chemokine production negative regulation of interferon-gamma production negative regulation of interleukin-1 beta production negative regulation of interleukin-10 production negative regulation of interleukin-2 production negative regulation of interleukin-5 production negative regulation of interleukin-6 production negative regulation of tumor necrosis factor production negative regulation of T cell proliferation negative regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of CD8-positive, alpha-beta T cell differentiation negative regulation of MAPK cascade negative regulation of cytotoxic T cell differentiation positive regulation of regulatory T cell differentiation negative regulation of osteoclast differentiation negative regulation of activated T cell proliferation negative regulation of T cell receptor signaling pathway negative regulation of IP-10 production interleukin-10-mediated signaling pathway negative regulation of cytokine production involved in inflammatory response negative regulation of protein localization to nucleus negative regulation of intracellular signal transduction negative regulation of pri-miRNA transcription from RNA polymerase II promoter negative regulation of T cell costimulation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-crr4t:80/100.66.76.64:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0