Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • INPP5E Antibodies

          Proteintech

          INPP5E Polyclonal Antibody

          50 References
          View all (11) INPP5E antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite INPP5E Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (23)
          INPP5E Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          INPP5E Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 23

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          INPP5E Antibody (17797-1-AP) in ICC/IF

          IF result of ciliary INPP5E in human hTERT-RPE1 cell: Upper is from control siRNA transfected cells and bottom is from INPP5E siRNA transfected cells. 17797-1-AP specifically recognizes the ciliary INPP5E (From Dr.Seongjin Seo). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          INPP5E Antibody in Immunocytochemistry (ICC/IF)
          INPP5E Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          INPP5E Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          INPP5E Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          INPP5E Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          INPP5E Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          INPP5E Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          INPP5E Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          INPP5E Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          INPP5E Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          INPP5E Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          INPP5E Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          INPP5E Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          INPP5E Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          INPP5E Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          INPP5E Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          INPP5E Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          INPP5E Antibody in Western Blot (WB)
          INPP5E Antibody in Western Blot (WB)
          INPP5E Antibody in Western Blot (WB)
          INPP5E Antibody in Western Blot (WB)
          INPP5E Antibody in Western Blot (WB)
          INPP5E Antibody in Immunoprecipitation (IP)
          INPP5E Polyclonal Antibody

          Product Details

          17797-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 14 publications 14 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 31 publications 31 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication

          Not applicable (N/A)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Zebrafish

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          INPP5E Fusion Protein Ag11959 (1-350 aa encoded by BC028032)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.36 mg/mL

          Amount

          54 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MPSKAENLR PSEPAPQPPE GRTLQGQLPG APPAQRAGSP PDAPGSESPA LACSTPATPS GEDPPARAAP IAPRPPARPR LERALSLDDK GWRRRRFRGS QEDLEARNGT SPSRGSVQSE GPGAPAHSCS PPCLSTSLQE IPKSRGVLSS ERGSPSSGGN PLSGVASSSP NLPHRDAAVA GSSPRLPSLL PPRPPPALSL DIASDSLRTA NKVDSDLADY KLRAQPLLVR AHSSLGPGRP RSPLACDDCS LRSAKSSFSL LAPIRSKDVR SRSYLEGSLL ASGALLGADE LARYFPDRNV ALFVATWNMQ GQKELPPSLD EFLLPAEADY AQDLYVIGVQ EGCSDRREWE T (1-350 aa encoded by BC028032)

          Target Information

          INPP5E (inositol polyphosphate 5-phosphatase), also known as phosphatidylinositol polyphosphate 5-phosphatase type IV, is a 644 amino acid peripheral membrane protein associated with Golgi stacks. Belonging to the inositol-1, 4, 5-trisphosphate 5-phosphatase type IV family, INPP5E converts phosphatidylinositol-3, 4, 5-triphosphate (PtdIns 3, 4, 5-P3) to PtdIns-P2. While inactive towards water soluble inositol phosphates, the activity of INPP5E is specific for lipid substrates. INPP5E becomes phosphorylated upon DNA damage and is expressed in brain, heart, pancreas, testis and spleen. Defects in INPP5E are the cause of Joubert syndrome type 1 (JBTS1), a disorder presenting with cerebellar ataxia, oculomotor apraxia, hypotonia, neonatal breathing abnormalities and psychomotor delay. In addition, mutations in the INPP5E gene may lead to mental retardation-truncal obesity-retinal dystrophy-micropenis (MORMS), an autosomal recessive disorder characterized by moderate mental retardation, truncal obesity, congenital non-progressive retinal dystrophy, and micropenis in males.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 5-phosphatase that induces arborization; 72 kDa inositol polyphosphate 5-phosphatase; inositol polyphosphate 5-phosphatase; Inositol polyphosphate-5-phosphatase E; inositol polyphosphate-5-phosphatase, 72 kDa; Pharbin; phosphatidylinositol (4,5) bisphosphate 5-phosphatase homolog; Phosphatidylinositol 4,5-bisphosphate 5-phosphatase; phosphatidylinositol 5-phosphatase; Phosphatidylinositol polyphosphate 5-phosphatase type IV; Phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase; phosphatidylinositol-4,5-bisphosphate 5-phosphatase

          View more View less

          Gene Aliases: 1200002L24Rik; 72kDa; CORS1; CPD4; INPP5E; JBTS1; mKIAA0123; MORMS; pharbin; PPI5PIV

          View more View less

          UniProt ID: (Human) Q9NRR6, (Rat) Q9WVR1, (Mouse) Q9JII1

          View more View less

          Entrez Gene ID: (Human) 56623, (Rat) 114089, (Mouse) 64436

          View more View less

          Function(s)
          phosphatidylinositol-4,5-bisphosphate 5-phosphatase activity inositol-polyphosphate 5-phosphatase activity protein binding phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase activity hydrolase activity phosphatase activity phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase activity phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity phosphatidylinositol-4,5-bisphosphate phosphatase activity phosphatase
          Process(es)
          lipid metabolic process phosphatidylinositol biosynthetic process phosphate-containing compound metabolic process negative regulation of translation organophosphate metabolic process phosphatidylinositol-3-phosphate biosynthetic process phosphatidylinositol metabolic process phosphatidylinositol dephosphorylation negative regulation of protein kinase B signaling cilium morphogenesis response to inositol negative regulation of protein localization to cilium positive regulation of neuron projection development inositol phosphate dephosphorylation dephosphorylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline