Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • IQGAP3 Antibodies

          Abnova

          IQGAP3 Polyclonal Antibody, MaxPab™

          View all (5) IQGAP3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite IQGAP3 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          IQGAP3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          IQGAP3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IQGAP3 Antibody (H00128239-D01P) in WB

          IQGAP3 MaxPab rabbit polyclonal antibody. Western Blot analysis of IQGAP3 expression in A-431. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IQGAP3 Antibody in Western Blot (WB)

          Product Details

          H00128239-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          IQGAP3 (NP_839943.2, 1 a.a. approximately 1631 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MERRAAGPGW AAYERLTAEE MDEQRRQNVA YQYLCRLEEA KRWMEACLKE ELPSPVELEE SLRNGVLLAK LGHCFAPSVV PLKKIYDVEQ LRYQATGLHF RHTDNINFWL SAIAHIGLPS TFFPETTDIY DKKNMPRVVY CIHALSLFLF RLGLAPQIHD LYGKVKFTAE ELSNMASELA KYGLQLPAFS KIGGILANEL SVDEAAVHAA VLAINEAVER GVVEDTLAAL QNPSALLENL REPLAAVYQE MLAQAKMEKA ANARNHDDRE SQDIYDHYLT QAEIQGNINH VNVHGALEVV DDALERQSPE ALLKALQDPA LALRGVRRDF ADWYLEQLNS DREQKAQELG LVELLEKEEV QAGVAAANTK GDQEQAMLHA VQRINKAIRR GVAADTVKEL MCPEAQLPPV YPVASSMYQL ELAVLQQQQG ELGQEELFVA VEMLSAVVLI NRALEARDAS GFWSSLVNPA TGLAEVEGEN AQRYFDALLK LRQERGMGED FLSWNDLQAT VSQVNAQTQE ETDRVLAVSL INEALDKGSP EKTLSALLLP AAGLDDVSLP VAPRYHLLLV AAKRQKAQVT GDPGAVLWLE EIRQGVVRAN QDTNTAQRMA LGVAAINQAI KEGKAAQTER VLRNPAVALR GVVPDCANGY QRALESAMAK KQRPADTAFW VQHDMKDGTA YYFHLQTFQG IWEQPPGCPL NTSHLTREEI QSAVTKVTAA YDRQQLWKAN VGFVIQLQAR LRGFLVRQKF AEHSHFLRTW LPAVIKIQAH WRGYRQRKIY LEWLQYFKAN LDAIIKIQAW ARMWAARRQY LRRLHYFQKN VNSIVKIQAF FRARKAQDDY RILVHAPHPP LSVVRRFAHL LNQSQQDFLA EAELLKLQEE VVRKIRSNQQ LEQDLNIMDI KIGLLVKNRI TLQEVVSHCK KLTKRNKEQL SDMMVLDKQK GLKSLSKEKR QKLEAYQHLF YLLQTQPIYL AKLIFQMPQN KTTKFMEAVI FSLYNYASSR REAYLLLQLF KTALQEEIKS KVEQPQDVVT GNPTVVRLVV RFYRNGRGQS ALQEILGKVI QDVLEDKVLS VHTDPVHLYK NWINQTEAQT GQRSHLPYDV TPEQALSHPE VQRRLDIALR NLLAMTDKFL LAITSSVDQI PYGMRYVAKV LKATLAEKFP DATDSEVYKV VGNLLYYRFL NPAVVAPDAF DIVAMAAGGA LAAPQRHALG AVAQLLQHAA AGKAFSGQSQ HLRVLNDYLE ETHLKFRKFI HRACQVPEPE ERFAVDEYSD MVAVAKPMVY ITVGELVNTH RLLLEHQDCI APDHQDPLHE LLEDLGELPT IPDLIGESIA ADGHTDLSKL EVSLTLTNKF EGLEADADDS NTRSLLLSTK QLLADIIQFH PGDTLKEILS LSASREQEAA HKQLMSRRQA CTAQTPEPLR RHRSLTAHSL LPLAEKQRRV LRNLRRLEAL GLVSARNGYQ GLVDELAKDI RNQHRHRHRR KAELVKLQAT LQGLSTKTTF YEEQGDYYSQ YIRACLDHLA PDSKSSGKGK KQPSLHYTAA QLLEKGVLVE IEDLPASHFR NVIFDITPGD EAGKFEVNAK FLGVDMERFQ LHYQDLLQLQ YEGVAVMKLF NKAKVNVNLL IFLLNKKFLR K

          Target Information

          IQGAP3 (IQ motif containing GTPase activating protein 3) is a 1,631 amino acid protein that acts as an effector of Cdc42 and Rac 1, linking their activation to the cytoskeleton during neuronal morphogenesis. A novel member of the IQGAP family, IQGAP3 is highly expressed in brain where it localizes to axons of hippocampal neurons. IQGAP3 contains one Ras-GAP domain, a CH (calponin-homology) domain, four IQ domains and is encoded by a gene located on human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome. Chromosome 1 houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson's disease, Gaucher disease, schizophrenia and Usher syndrome. Aberrations in chromosome 1 are found in a variety of cancers, including head and neck cancer, malignant melanoma and multiple myeloma.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: IQGA3; IQGAP3; MGC10170; Ras GTPase-activating-like protein IQGAP3; unnamed protein product

          View more View less

          Gene Aliases: IQGAP3

          View more View less

          UniProt ID: (Human) Q86VI3

          View more View less

          Entrez Gene ID: (Human) 128239

          View more View less

          Function(s)
          GTPase activator activity protein binding calmodulin binding small GTPase binding actin filament binding myosin VI light chain binding
          Process(es)
          G1/S transition of mitotic cell cycle Ras protein signal transduction regulation of cell size gene expression positive regulation of gene expression negative regulation of gene expression mammary gland epithelial cell proliferation positive regulation of mammary gland epithelial cell proliferation positive regulation of MAPK cascade ERK1 and ERK2 cascade mitotic actomyosin contractile ring assembly actin filament organization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline