Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • IQSEC2 Antibodies

          Abnova

          IQSEC2 Polyclonal Antibody, MaxPab™

          View all (4) IQSEC2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite IQSEC2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          IQSEC2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          IQSEC2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IQSEC2 Antibody (H00023096-B01P) in WB

          Western Blot analysis of IQSEC2 expression in transfected 293T cell line by IQSEC2 MaxPab polyclonal antibody. Lane 1: IQSEC2 transfected lysate(104.39 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IQSEC2 Antibody in Western Blot (WB)
          IQSEC2 Polyclonal Antibody, MaxPab™

          Product Details

          H00023096-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          IQSEC2 (AAI56684.1, 1 a.a. approximately 949 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.46 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MEPPGRSSRS TASHTLHQYC CPTQVLDSMK LTPSGRLAES SVEGDAPGSD LSTAVDSPGS QPPYRLSQLP PSSSHMGGPP AGVGLPWAQR ARLQPASVAL RKQEEEEIKR SKALSDSYEL STDLQDKKVE MLERKYGGSF LSRRAARTIQ TAFRQYRMNK NFERLRSSAS ESRMSRRIIL SNMRMQFSFE EYEKAQNPAY FEGKPASLDE GAMAGARSHR LERGLPYGGS CGGGIDGGGS SVTTSGEFSN DITELEDSFS KQVKSLAESI DEALNCHPSG PMSEEPGSAQ LEKRESKEQQ EDSSATSFSD LPLYLDDTVP QQSPERLPST EPPPQGRPEF WAPAPLPPVP PPVPSGTRED GSREEGTRRG PGCLECRDFR LRAAHLPLLT IEPPSDSSVD LSDRSDRGSV HRQLVYEADG CSPHGTLKHK GPPGRAPIPH RHYPAPEGPA PAPPGPLPPA PNSGTGPSGV AGGRRLGKCE AAGENSDGGD NESLESSSNS NETINCSSGS SSRDSLREPP ATGLCKQTYQ RETRHSWDSP AFNNDVVQRR HYRIGLNLFN KKPEKGIQYL IERGFLSDTP VGVAHFILER KGLSRQMIGE FLGNRQKQFN RDVLDCVVDE MDFSSMDLDD ALRKFQSHIR VQGEAQKVER LIEAFSQRYC VCNPALVRQF RNPDTIFILA FAIILLNTDM YSPSVKAERK MKLDDFIKNL RGVDNGEDIP RDLLVGIYQR IQGRELRTND DHVSQVQAVE RMIVGKKPVL SLPHRRLVCC CQLYEVPDPN RPQRLGLHQR EVFLFNDLLV VTKIFQKKKI LVTYSFRQSF PLVEMHMQLF QNSYYQFGIK LLSAVPGGER KVLIIFNAPS LQDRLRFTSD LRESIAEVQE MEKYRVESEL EKQKGMMRPN ASQPGGAKDS VNGTMARSSL EDTYGAGDGL KRGALSSSLR DLSDAGVCY

          Target Information

          This gene encodes a guanine nucleotide exchange factor for the ARF family of small GTP-binding proteins. The encoded protein is a component of the postsynaptic density at excitatory synapses, and may play a critical role in cytoskeletal and synaptic organization through the activation of selected ARF substrates including ARF1 and ARF6. Mutations in this gene have been implicated in nonsyndromic X-linked mental retardation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: BRAG1b; IQsec2; BRAG1c; IQSec2; brefeldin A resistant Arf-guanine nucleotide exchange factor 1; brefeldin A resistant Arf-guanine nucleotide exchange factor 1b; brefeldin A resistant Arf-guanine nucleotide exchange factor 1c; IQ motif and Sec7 domain 2; IQ motif and SEC7 domain-containing protein 2; IQsec2; unnamed protein product

          View more View less

          Gene Aliases: BRAG1; IQ-ArfGEF; IQSEC2; KIAA0522; MRX1; MRX18; MRX78; XLID1

          View more View less

          UniProt ID: (Human) Q5JU85

          View more View less

          Entrez Gene ID: (Human) 23096

          View more View less

          Function(s)
          guanyl-nucleotide exchange factor activity protein binding
          Process(es)
          actin cytoskeleton organization regulation of ARF protein signal transduction modulation of synaptic transmission positive regulation of synapse assembly positive regulation of synaptic transmission, glutamatergic regulation of neurotransmitter receptor localization to postsynaptic specialization membrane positive regulation of long term synaptic depression
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline