Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MNQLSFLLF LIATTRGWST DEANTYFKEW TCSSSPSLPR SCKEIKDECP SAFDGLYFLR TENGVIYQTF CDMTSGGGGW TLVASVHEND MRGKCTVGDR WSSQQGSKAV YPEGDGNWAN YNTFGSAEAA TSDDYKNPGY YDIQAKDLGI WHVPNKSPMQ HWRNSSLLRY RTDTGFLQTL GHNLFGIYQK YPVKYGEGKC WTDNGPVIPV VYDFGDAQKT ASYYSPYGQR EFTAGFVQFR VFNNERAANA LCAGMRVTGC NTEHHCIGGG GYFPEASPQQ CGDFSGFDWS GYGTHVGYSS SREITEAAVL LFYR (1-313 aa encoded by BC020664)
ITLN1 has no effect on basal glucose uptake but enhances insulin-stimulated glucose uptake in adipocytes. ITLN1 increases AKT phosphorylation in the absence and presence of insulin. ITLN1 may play a role in the defense system against microorganisms. ITLN1 may specifically recognize carbohydrate chains of pathogens and bacterial components containing galactofuranosyl residues, in a calcium-dependent manner. ITLN1 may be involved in iron metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium-dependent galactosyl-specific lectin; calcium-dependent galactosyl-specific lectin; LFR; HL-1; ITLN; hIntL; FLJ20022; endothelial lectin HL-1; FLJ20022; galactofuranose-binding lectin; intelectin 1 (galactofuranose binding); intelectin 1 (galactofuranose binding)-like; intelectin 2; intelectin 5; intelectin a; intestinal lactoferrin receptor; ITLN-1; ITLN1; unnamed protein product
Gene Aliases: hIntL; HL-1; HL1; INTL; ITLN; Itln2; Itln3; Itln5; Itlna; LFR; omentin
UniProt ID: (Human) Q8WWA0, (Mouse) O88310
Entrez Gene ID: (Human) 55600, (Mouse) 16429, (Rat) 498284
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support