Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • ITPKB Antibodies

          Abnova

          ITPKB Polyclonal Antibody, MaxPab™

          View all (9) ITPKB antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ITPKB Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          ITPKB Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ITPKB Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ITPKB Antibody (H00003707-B01P) in WB

          Western Blot analysis of ITPKB expression in transfected 293T cell line by ITPKB MaxPab polyclonal antibody. Lane1:ITPKB transfected lysate(70.84 KDa). Lane2:Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ITPKB Antibody in Western Blot (WB)

          Product Details

          H00003707-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ITPKB (AAH15009.1, 1 a.a. approximately 644 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAVYCYALNS LVIMNSANEM KSGGGPGPSG SETPPPPRRA VLSPGSVFSP GRGASFLFPP AESLSPEEPR SPGGWRSGRR RLNSSSGSGS GSSGSSVSSP SWAGRLRGDR QQVVAAGTLS PPGPEEAKRK LRILQRELQN VQVNQKVGMF EAHIQAQSSA IQAPRSPRLG RAHSPSPCPF RSSSQPPGRV LVQGARSEER RTKSWGEQCP ETSGTDSGRK GGPSLCSSQV KKGMPPLPGR AAPTGSEAQG PSAFVRMEKG IPASPRCGSP TAMEIDKRGS PTPGTRSCLA PSLGLFGASL TMATEVAARV TSTGPHRPQD LALTEPSGRA RELEDLQPPE ALVERQGQFL GSETSPAPER GGPRDGEPPG KMGKGYLPCG MPGSGEPEVG KRPEETTVSV QSAESSDALS WSRLPRALAS VGPEEARSGA PVGGGRWQLS DRVEGGSPTL GLLGGSPSAQ PGTGNVEAGI PSGRMLEPLP CWDAAKDLKE PQCPPGDRVG VQPGNSRVWQ GTMEKAGLAW TRGTGVQSEG TWESQRQDSD ALPSPELLPQ DQDKPFLRKA CSPSNIPAVI ITDMGTQEDG ALEETQGSPR GNLPLRKLSS SSASSTGFSS SYEDSEEDIS SDPERTLDPN SAFLHTLDQQ KPRV

          Target Information

          Inositol 1,4,5-trisphosphate (Ins(1,4,5)P3) regulates the level of calcium within the cell by releasing calcium from intracellular stores. Ins(1,4,5)P3 is phosphorylated by inositol 1,4,5-trisphosphate 3-kinase (IP3K) to form inositol 1,3,4,5-tetrakisphosphate (Ins(1,4,5)P4), which is is thought to regulate the influx of calcium across the plasma membrane. IP3K exists as three isoforms, IP3KA, B, and C. IP3KA, the most highly characterized isoform, is expressed in rat brain and testis. IP3KB is expressed in various rat tissues such as lung, thymus, testis, brain, and heart. IP3K activity is stimulated in the presence of calmodulin via phosphorylation by cAMP-dependent protein kinase, protein kinase C, or calcium/calmodulin dependent protein kinase II and, subsequently, mediates the inositol phosphate signaling pathways.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Inositol 1,4,5-trisphosphate 3-kinase B; Inositol-trisphosphate 3-kinase B; insP 3-kinase B; IP3 3-kinase B; IP3K B; proliferation-inducing protein 37; unnamed protein product

          View more View less

          Gene Aliases: IP3-3KB; IP3K; IP3K-B; IP3KB; ITPKB; PIG37

          View more View less

          UniProt ID: (Human) P27987

          View more View less

          Entrez Gene ID: (Human) 3707

          View more View less

          Function(s)
          nucleotide binding inositol hexakisphosphate kinase activity protein binding calmodulin binding ATP binding inositol-1,4,5-trisphosphate 3-kinase activity kinase activity transferase activity
          Process(es)
          MAPK cascade myeloid cell homeostasis signal transduction cell surface receptor signaling pathway T cell differentiation inositol trisphosphate metabolic process inositol phosphate biosynthetic process negative regulation of neutrophil apoptotic process common myeloid progenitor cell proliferation positive thymic T cell selection thymic T cell selection negative regulation of myeloid cell differentiation positive regulation of Ras protein signal transduction positive regulation of alpha-beta T cell differentiation phosphatidylinositol phosphorylation cellular response to calcium ion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline