Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: PVTSYNWLTD FPSQEALDSF YKHYVQNAIR DLPCTCEFVE SFVDDLIEAC RVLSRQEAHP QLEDCLGIGA AFEKWGTLHE TQKFDILVPI VPPQGTMFVL EMRDPALGRR CGCVLVESEC VCKREKLLGD VL
Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.
The ITPRIPL1 (Inositol 1,4,5-Trisphosphate Receptor Interacting Protein-Like 1) gene encodes a protein implicated in T cell regulation by interacting with the CD3ε molecule. It is located on chromosome 6. The ITPRIPL1 protein acts as an inhibitory ligand for CD3ε, thereby modulating T cell activation in the tumor microenvironment. This interaction significantly reduces calcium influx and phosphorylation of ZAP70, which are critical steps in the T cell activation pathway. The downregulation of these processes by ITPRIPL1 suggests its role in immune response modulation and tumor immune evasion. Given its regulatory functions, ITPRIPL1 has become a subject of research for its potential implications in cancer immunotherapy and as a biomarker for diagnosing and prognosticating various cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support