Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 9
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
MAPK9 (JNK2) is a serine/threonine kinase which plays an important role in the MAPK pathway. JNK2 (p54a, SAPK1a), along with JNK1 and JNK3, is thought to play an important role in nuclear signal transduction through its environmental stress activation and subsequent phosphorylation of the nuclear transcription factor p53.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-Jun kinase 2; c-Jun N-terminal kinase 2; JNK; JNK-55; JNK/SAPK alpha; JNK1; JNK1A2; JNK21B1/2; Jun kinase; MAP kinase 9; MAPK 10; MAPK 9; MAPK10; MAPK8; mitogen activated protein kinase 9; Mitogen-activated protein kinase 9; OTTHUMP00000161542; OTTHUMP00000161543; p54-alpha; PRKM8; protein kinase, mitogen-activated 9; SAPK-alpha; SAPK1; SAPK1a; stress activated protein kinase alpha II; Stress-activated protein kinase 1a; Stress-activated protein kinase JNK2
Gene Aliases: AI851083; JNK-55; JNK2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; MAPK9; p54a; p54aSAPK; PRKM9; SAPK; SAPK1A
UniProt ID: (Human) P45984, (Rat) P49186, (Mouse) Q9WTU6
Entrez Gene ID: (Human) 5601, (Rat) 50658, (Mouse) 26420
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support