Search
Search
Rockland
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
A BLAST analysis was used to suggest cross-reactivity with human, rat, and mouse based on 100% sequence homology. Cross-reactivity with KAT II from other sources has not been determined.
This gene encodes a protein that is highly similar to mouse and rat kynurenine aminotransferase II. The rat protein is a homodimer with two transaminase activities. One activity is the transamination of alpha-aminoadipic acid, a final step in the saccaropine pathway which is the major pathway for L-lysine catabolism. The other activity involves the transamination of kynurenine to produce kynurenine acid, the precursor of kynurenic acid which has neuroprotective properties. Two alternative transcripts encoding the same isoform have been identified, however, additional alternative transcripts and isoforms may exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-aminoadipate aminotransferase; 2-aminoadipate transaminase; alpha-aminoadipate aminotransferase; glycine transaminase AADAT; KAT/AadAT; kynurenine aminotransferase II; kynurenine--glyoxylate transaminase AADAT; kynurenine--oxoglutarate; kynurenine--oxoglutarate aminotransferase II; kynurenine--oxoglutarate transaminase 2; kynurenine--oxoglutarate transaminase II; methionine--glyoxylate transaminase AADAT; unnamed protein product
Gene Aliases: Aadt; Kat2; KATII; Kyat2; mKat-2
UniProt ID: (Mouse) Q9WVM8
Entrez Gene ID: (Mouse) 23923
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support