Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • KCNK2 Antibodies

          Osenses

          KCNK2 Polyclonal Antibody

          1 Reference
          View all (6) KCNK2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite KCNK2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          OSK00038W-100UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (IHC)

          1:1,000
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Rabbit / Ig

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide from human KCNK2 conjugated to blue carrier protein was used as the antigen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.

          The peptide is homologous in rat and mouse.

          Specificity of this antibody: KCNK2.

          Target Information

          TREK-1 is a two-pore-domain background potassium channel expressed throughout the central nervous system. It is opened by polyunsaturated fatty acids and lysophospholipids. As well, it is regulated by various neurotransmitters. It has been shown that alterations in the functioning, regulation, or both of the TREK-1 channel may alter mood. TREK-1 is also activated by volatile anesthetics and has been suggested to be an important target in the action of these drugs. Therefore, this particular K+ channel emerges as a potential innovative target for developing new therapeutic agents for anesthesiology and neurology, such as antidepressants.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: arachidonic acid sensitive tandem pore domain potassium channel; hTREK-1c; hTREK-1e; ion transport membrane protein; K2P2.1; K2P2.1 potassium channel; kcnk2 {ECO:0000312|EMBL:AAU06141.1, ECO:0000312|Ensembl:ENSRNOP00000003684, ECO:0000312|RGD:621448}; Outward rectifying potassium channel protein TREK-1; outward rectifying potassium channel protein TREK-1 {ECO:0000250|UniProtKB:O95069}; Potassium channel subfamily K member 2; potassium channel subfamily K member 2 {ECO:0000250|UniProtKB:O95069}; potassium channel, subfamily K, member 2; potassium channel, two pore domain subfamily K, member 2; potassium inwardly-rectifying channel, subfamily K, member 2; Stretch-activated potassium channel TREK-1; stretch-activated potassium channel TREK-1 {ECO:0000303|PubMed:16248991}; tandem-pore-domain potassium channel TREK-1; trek 1; TREK-1 K(+) channel subunit; TREK-1 K(+) channel subunit {ECO:0000250|UniProtKB:O95069}; TWIK-related potassium channel 1; two pore domain K+ channel subunit; Two pore domain potassium channel TREK-1; two pore domain potassium channel TREK-1 {ECO:0000303|PubMed:24196565}; Two pore domain potassium channel TREK1; Two pore potassium channel TPKC1; two pore potassium channel TPKC1 {ECO:0000250|UniProtKB:O95069}; two-pore potassium channel 1; two-pore-domain potassium channel subunit; unnamed protein product

          View more View less

          Gene Aliases: A430027H14Rik; AI848635; hTREK-1c; hTREK-1e; K2p2.1; KCNK2; rTREK1d; TPKC1; TREK; TREK-1; TREK1

          View more View less

          UniProt ID: (Human) O95069, (Rat) Q920B6, (Mouse) P97438

          View more View less

          Entrez Gene ID: (Human) 3776, (Rat) 170899, (Mouse) 16526

          View more View less

          Function(s)
          voltage-gated potassium channel activity potassium channel activity outward rectifier potassium channel activity potassium channel inhibitor activity ligand-gated channel activity potassium ion leak channel activity identical protein binding metal ion binding protein heterodimerization activity mechanically-gated potassium channel activity protein binding voltage-gated ion channel activity ion channel transporter
          Process(es)
          cardiac ventricle development ion transport potassium ion transport G-protein coupled receptor signaling pathway memory response to mechanical stimulus glutamate secretion neuronal action potential regulation of synaptic transmission, GABAergic ion transmembrane transport regulation of membrane potential response to axon injury detection of mechanical stimulus involved in sensory perception of touch negative regulation of cardiac muscle cell proliferation cellular response to hypoxia potassium ion transmembrane transport cochlea development chemical synaptic transmission, postsynaptic positive regulation of cellular response to hypoxia chloride transmembrane transport cellular response to arachidonate negative regulation of DNA biosynthetic process transport positive regulation of cell death stabilization of membrane potential regulation of ion transmembrane transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline