Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • KCNQ2 Antibodies

          Invitrogen

          KCNQ2 Polyclonal Antibody

          7 Published Figures
          9 References
          View all (9) KCNQ2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite KCNQ2 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          • Published Figures (7)
          KCNQ2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          KCNQ2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          KCNQ2 Antibody (PA1-929) in ICC/IF

          Immunofluorescent analysis of KCNQ2 using Anti-KCNQ2 Polyclonal Antibody (Product # PA1-929) shows staining in HepG2 Cells. KCNQ2 staining (green), F-Actin staining with Phalloidin (red) and nuclei with DAPI (blue) is shown. Cells were grown on chamber slides and fixed with formaldehyde prior to staining. Cells were probed without (control) or with or an antibody recognizing KCNQ2 (Product # PA1-929) at a dilution of 1:100 over night at 4°C, washed with PBS and incubated with a DyLight-488 conjugated secondary antibody (Product # 35552, Go... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          KCNQ2 Antibody in Immunocytochemistry (ICC/IF)
          KCNQ2 Antibody in Immunocytochemistry (ICC/IF)
          KCNQ2 Antibody in Immunocytochemistry (ICC/IF)
          KCNQ2 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          KCNQ2 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          KCNQ2 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          KCNQ2 Antibody in Immunocytochemistry (ICC/IF)
          KCNQ2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KCNQ2 Antibody in Western Blot (WB)
          KCNQ2 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          KCNQ2 Polyclonal Antibody

          Product Details

          PA1-929

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          0.5 µg/mL
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          0.5 µg/mL
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Gerbil, Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          GST fusion protein encoding the first 70 amino acids of human KCNQ2.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2131689

          Product Specific Information

          PA1-929 detects KCNQ2 from mouse, human and rat samples. This antibody is specific for KCNQ2 and does not detect KCNQ1, KCNQ3, KCNQ4 or KCNQ5.

          PA1-929 has been successfully used in Western blot, immunofluorescence, immunohistochemistry and immunocytochemistry procedures. Immunohistochemical staining of rat hippocampal using PA1-929 yielded a strong signal in granule cell layer and the mossy fibers found in the central hilus of the dentate gyrus.

          PA1-929 immunogen is a GST fusion protein encoding the first 70 amino acids of human KCNQ2.

          Target Information

          KCNQs are members of the voltage-dependent non-inactivating potassium channel family. Currently there are five known KNCQs (KCNQ1-5) found in the central nervous system and KNCQ2 and 3 have demonstrated their importance in M-current activation. Studies have shown that KCNQ2 and KCNQ3 form heteromultimers that, when formed, substantially increase the M-current. Inhibition of M-current controls neuron excitability throughout the nervous system as well as the responsiveness to synaptic inputs. Genetic mutations in these proteins have been linked to disorders such as benign familial neonatal convulsions (BFNC), deafness, neuropathic pain and epilepsy.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: KQT like 2; KQT-like 2; KVEBN1; neuroblastoma-specific potassium channel subunit alpha KvLQT2; potassium channel subunit alpha KvLQT2; potassium channel, voltage gated KQT-like subfamily Q, member 2; potassium channel, voltage-gated KQT-like subfamily Q, member 2; potassium voltage-gated channel, KQT-like subfamily, member 2; unnamed protein product; voltage-gated potassium channel subunit Kv7.2

          View more View less

          Gene Aliases: BFNC; DEE7; EBN; EBN1; ENB1; HNSPC; KCNA11; KQT2; KV7.2; Nmf134

          View more View less

          UniProt ID: (Human) O43526, (Rat) O88943, (Mouse) Q9Z351

          View more View less

          Entrez Gene ID: (Human) 3785, (Rat) 170848, (Mouse) 16536

          View more View less

          Function(s)
          ion channel activity voltage-gated potassium channel activity potassium channel activity protein binding calmodulin binding voltage-gated cation channel activity ankyrin binding voltage-gated ion channel activity
          Process(es)
          action potential protein phosphorylation protein import into nucleus ion transport potassium ion transport exocytosis endocytosis brain development sensory perception of sound learning memory grooming behavior determination of adult lifespan response to temperature stimulus response to xenobiotic stimulus response to mechanical stimulus gene expression response to auditory stimulus protein transport neuron remodeling transmission of nerve impulse neuronal action potential dentate gyrus development nerve development hippocampus development establishment of cell polarity neuron differentiation ion transmembrane transport social behavior exploration behavior sodium ion transmembrane transport psychomotor behavior regulation of membrane potential regulation of synaptic plasticity cognition neuron apoptotic process mitochondrial depolarization transmembrane transport motor behavior cellular response to calcium ion cellular response to xenobiotic stimulus potassium ion transmembrane transport apoptosome assembly hippocampal pyramidal neuron differentiation excitatory chemical synaptic transmission inhibitory chemical synaptic transmission action potential initiation regulation of action potential firing threshold substantia propria of cornea development chemical synaptic transmission nervous system development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-kp6hx:80/100.66.72.150:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0