Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: DEEAVDREP RRLSSRRSVL TSPVANGVNL DYDGLGKTCR SLPSLKKTLA GDSSSDCSRG SHNGQVWDPQ CAPRKDRQVH LTHFELEGLR CLVDKLESLP LHKKCVPTGI EDEDALIADV KILLEELANS DPKLALTGVP IVQWPKRDKL KFPTRPKVRV PTIPITKPHT MKPAPRLTPV RPAAAS (374-558 aa encoded by BC064360)
FBXL11 is a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box). The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. FBXL11 belongs to the Fbls class and, in addition to an F-box, contains at least 6 highly degenerated leucine-rich repeats.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [Histone-H3]-lysine-36 demethylase 1A; CXXC-type zinc finger protein 8; DKFZp434M1735; F-box and leucine-rich repeat protein 11; F-box protein containing leucine repeats; F-box/LRR-repeat protein 11; FLJ00115; FLJ46431; jmjC domain-containing histone demethylation protein 1A; jumonji C domain-containing histone demethylase 1A; KDM2A-C620; KDM2A-delta N; KDM2A-N782; lysine (K)-specific demethylase 2A; SF-KDM2A; unnamed protein product
Gene Aliases: 100043628; 5530401A10Rik; CXXC8; FBL11; FBL7; FBXL11; Gm4560; Jhdm1; JHDM1A; lalina; LILINA
UniProt ID: (Human) Q9Y2K7, (Mouse) P59997
Entrez Gene ID: (Human) 22992, (Mouse) 225876
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support