Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: SRQNKSVLR PAVTNGMSQL PSINPSASSG NETTFSGGGG PAPVTTPEPD HVPKADSTDI RSEEPLKTDS SASNSNSELK AIRPPCPDTA PPSSALHWLA DLATQKAKEE TKEAGSLRSV LNKESHSPFG LDSFNSTAKV SPLTPKLFNS LLLGPTASNN KTEGSSLRDL LHSGPGKLPQ TPLDTGIPFP PVFSTSSAGV KSKASLPNFL DHIIASVVEN KKTSDASKRA CNLTDTQKEV KEMVMGLNVL DPHTSHS (1127-1382 aa encoded by BC001202)
Histone demethylase that specifically demethylates 'Lys-9' of histone H3, thereby playing a central role in histone code. Demethylation of Lys residue generates formaldehyde and succinate. May have tumor suppressor activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [histone H3]-dimethyl-L-lysine(9) demethylase 3B; jmjC domain-containing histone demethylation protein 2B; jumonji domain containing 1B; jumonji domain-containing protein 1B; lysine (K)-specific demethylase 3B; nuclear protein 5qNCA; PZP; unnamed protein product
Gene Aliases: 5830462I21Rik; 5qNCA; C5orf7; DIJOS; JHDM2B; JMJD1B; mKIAA1082; NET22
UniProt ID: (Human) Q7LBC6, (Mouse) Q6ZPY7
Entrez Gene ID: (Human) 51780, (Mouse) 277250
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support