Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • KIF3B Antibodies

          Abnova

          KIF3B Polyclonal Antibody, MaxPab™

          View all (5) KIF3B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite KIF3B Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          KIF3B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          KIF3B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          KIF3B Antibody (H00009371-B01P) in WB

          KIF3B MaxPab polyclonal antibody. Western Blot analysis of KIF3B expression in HeLa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          KIF3B Antibody in Western Blot (WB)
          KIF3B Polyclonal Antibody, MaxPab™

          Product Details

          H00009371-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Rat

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KIF3B (ENSP00000364864, 1 a.a. approximately 747 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSKLKSSESV RVVVRCRPMN GKEKAASYDK VVDVDVKLGQ VSVKNPKGTA HEMPKTFTFD AVYDWNAKQF ELYDETFRPL VDSVLQGFNG TIFAYGQTGT GKTYTMEGIR GDPEKRGVIP NSFDHIFTHI SRSQNQQYLV RASYLEIYQE EIRDLLSKDQ TKRLELKERP DTGVYVKDLS SFVTKSVKEI EHVMNVGNQN RSVGATNMNE HSSRSHAIFV ITIECSEVGL DGENHIRVGK LNLVDLAGSE RQAKTGAQGE RLKEATKINL SLSALGNVIS ALVDGKSTHI PYRDSKLTRL LQDSLGGNAK TVMVANVGPA SYNVEETLTT LRYANRAKNI KNKPRVNEDP KDALLREFQE EIARLKAQLE KRSIGRRKRR EKRREGGGSG GGGEEEEEEG EEGEEEGDDK DDYWREQQEK LEIEKRAIVE DHSLVAEEKM RLLKEKEKKM EDLRREKDAA EMLGAKIKAM ESKLLVGGKN IVDHTNEQQK ILEQKRQEIA EQKRREREIQ QQMESRDEET LELKETYSSL QQEVDIKTKK LKKLFSKLQA VKAEIHDLQE EHIKERQELE QTQNELTREL KLKHLIIENF IPLEEKSKIM NRAFFDEEED HWKLHPITRL ENQQMMKRPV SAVGYKRPLS QHARMSMMIR PEARYRAENI VLLELDMPSR TTRDYEGPAI APKVQAALDA ALQDEDEIQV DASSFESTAN KKSKARPKSG RKSGSSSSSS GTPASQLYPQ SRGLVPK

          Target Information

          The PARP11 gene is part of the poly (ADP-ribose) polymerase family.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: HH0048; KIAA0359; Kinesin-like protein KIF3B; Microtubule plus end-directed kinesin motor 3B; unnamed protein product

          View more View less

          Gene Aliases: FLA8; HH0048; KIAA0359; KIF3B; KLP-11; OTSC12; RP89

          View more View less

          UniProt ID: (Human) O15066

          View more View less

          Entrez Gene ID: (Human) 9371, (Rat) 296284

          View more View less

          Function(s)
          nucleotide binding microtubule motor activity protein binding ATP binding microtubule binding ATP-dependent microtubule motor activity, plus-end-directed ATPase activity small GTPase binding intraciliary transport particle B binding Rho GTPase binding microtubule binding motor protein
          Process(es)
          microtubule-based movement mitotic spindle organization mitotic centrosome separation determination of left/right symmetry axo-dendritic transport anterograde axonal transport vesicle-mediated transport protein complex localization positive regulation of cytokinesis opsin transport intraciliary transport cilium morphogenesis plus-end-directed vesicle transport along microtubule mitotic spindle assembly anterograde dendritic transport of neurotransmitter receptor complex metabolic process cytoskeleton-dependent intracellular transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline