Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASKTKASEA LKVVARCRPL SRKEEAAGHE QILTMDVKLG QVTLRNPRAA PGELPKTFTF DAVYDASSKQ ADLYDETVRP LIDSVLQGFN GTVFAYGQTG TGKTYTMQGT WVEPELRGVI PNAFEHIFTH ISRSQNQQYL VRASYLEIYQ EEIRDLLSKE PGKRLELKEN PETGVYIKDL SSFVTKNVKE IEHVMNLGNQ TRAVGSTHMN EVSSRSHAIF IITVECSERG SDGQDHIRVG KLNLVDLAGS ERQNKAGPNT AGGAATPSSG GGGGGGGSGG GAGGERPKEA SKINLSLSAL GNVIAALAGN RSTHIPYRDS KLTRLLQDSL GGNAKTIMVA TLGPASHSYD ESLSTLRFAN RAKNIKNKPR VNEDPKDTLL REFQEEIARL KAQLEKRGML GKRPRRKSSR RKKAVSAPPG YPEGPVIEAW VAEEEDDNNN NHRPPQPILE SALEKNMENY LQEQKERLEE EKAAIQDDRS LVSEEKQKLL EEKEKMLEDL RREQQATELL AAKYKAMESK LLIGGRNIMD HTNEQQKMLE LKRQEIAEQK RREREMQQEM MLRDEETMEL RGTYTSLQQE VEVKTKKLKK LYAKLQAVKA EIQDQHDEYI RVRQDLEEAQ NEQTRELKLK YLIIENFIPP EEKNKIMNRL FLDCEEEQWK FQPLVPAGVS SSQMKKRPTS AVGYKRPISQ YARVAMAMGS HPRYRAENIM FLELDVSPPA VFEMEFSHDQ EQDPRALHME RLMRLDSFLE RPSTSKVRKS RSWCQSPQRP PPSTTHASLA SASLRPATVA DHE
KIF3C is a microtubule-based anterograde translocator for membranous organelles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KIF3C variant protein; Kinesin-like protein KIF3C; unnamed protein product
Gene Aliases: KIF3C
UniProt ID: (Human) O14782
Entrez Gene ID: (Human) 3797
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support