Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • KMO Antibodies

          Proteintech

          KMO Polyclonal Antibody

          28 References
          View all (15) KMO antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite KMO Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (21)
          KMO Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          KMO Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 21

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          KMO Antibody (10698-1-AP) in ICC/IF

          Immunofluorescent analysis of (-20°C Ethanol) fixed MCF-7 cells using 10698-1-AP (KMO antibody) at dilution of 1:50 and Alexa Fluor 488-Conjugated AffiniPure Goat Anti-Rabbit IgG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          KMO Antibody in Immunocytochemistry (ICC/IF)
          KMO Antibody in Immunocytochemistry (ICC/IF)
          KMO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KMO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KMO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KMO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KMO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KMO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KMO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KMO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KMO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KMO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KMO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KMO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KMO Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KMO Antibody in Western Blot (WB)
          KMO Antibody in Western Blot (WB)
          KMO Antibody in Western Blot (WB)
          KMO Antibody in Western Blot (WB)
          KMO Antibody in Western Blot (WB)
          KMO Antibody in Western Blot (WB)
          KMO Polyclonal Antibody

          Product Details

          10698-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 12 publications 12 publications

          Immunohistochemistry (IHC)

          -
          View 8 publications 8 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:400
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 8 publications 8 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KMO Fusion Protein Ag1109 (107-407 aa encoded by BC005297)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.26 mg/mL

          Amount

          39 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: LSVSRENLN KDLLTAAEKY PNVKMHFNHR LLKCNPEEGM ITVLGSDKVP KDVTCDLIVG CDGAYSTVRS HLMKKPRFDY SQQYIPHGYM ELTIPPKNGD YAMEPNYLHI WPRNTFMMIA LPNMNKSFTC TLFMPFEEFE KLLTSNDVVD FFQKYFPDAI PLIGEKLLVQ DFFLLPAQPM ISVKCSSFHF KSHCVLLGDA AHAIVPFFGQ GMNAGFEDCL VFDELMDKFS NDLSLCLPVF SRLRIPDDHA ISDLSMYNYI EKNMERFLHA IMPSTFIPLY TMVTFSRIRY HEAVQRWHWQ KR (107-407 aa encoded by BC005297)

          Target Information

          Kynurenine 3-monooxygenase (KMO; EC 1.14.13.9) is an NADPH-dependent flavin monooxygenase that catalyzes the hydroxylation of the L-tryptophan metabolite L-kynurenine to form L-3-hydroxykynurenine.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: hK3OH-2; kmo; kmo {ECO:0000312|RGD:620610}; Kynurenine 3-hydroxylase; kynurenine 3-hydroxylase {ECO:0000255|HAMAP-Rule:MF_03018}; Kynurenine 3-monooxygenase; kynurenine 3-monooxygenase {ECO:0000255|HAMAP-Rule:MF_03018}; unnamed protein product

          View more View less

          Gene Aliases: AI046660; dJ317G22.1; KMO

          View more View less

          UniProt ID: (Human) O15229, (Mouse) Q91WN4, (Rat) O88867

          View more View less

          Entrez Gene ID: (Human) 8564, (Mouse) 98256, (Rat) 59113

          View more View less

          Function(s)
          monooxygenase activity kynurenine 3-monooxygenase activity protein binding NAD(P)H oxidase activity oxidoreductase activity flavin adenine dinucleotide binding FAD binding oxygenase
          Process(es)
          tryptophan catabolic process NAD biosynthetic process response to salt stress positive regulation of glutamate secretion pyridine nucleotide biosynthetic process NAD metabolic process quinolinate biosynthetic process response to lipopolysaccharide kynurenic acid biosynthetic process 'de novo' NAD biosynthetic process from tryptophan anthranilate metabolic process kynurenine metabolic process cellular response to lipopolysaccharide cellular response to interleukin-1 L-kynurenine metabolic process positive regulation of glutamate secretion, neurotransmission metabolic process tryptophan catabolic process to kynurenine secondary metabolite biosynthetic process oxidation-reduction process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-d4j86:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline