Search Thermo Fisher Scientific
Mediates the voltage-dependent potassium ion permeability of excitable membranes. Channels open or close in response to the voltage difference across the membrane, letting potassium ions pass in accordance with their electrochemical gradient.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Delayed rectifier potassium channel 1; DRK1; KCB1; mShab; potassium channel Kv2.1; potassium voltage gated channel, Shab-related subfamily, member 1; Potassium voltage-gated channel subfamily B member 1; potassium voltage-gated channel, Shab-related subfamily, member 1; Voltage-gated potassium channel subunit Kv2.1
Gene Aliases: DRK1; DRK1PC; KCNB1; Kcr1-1; Kv2.1; Shab
UniProt ID: (Human) Q14721, (Mouse) Q03717, (Rat) P15387
Entrez Gene ID: (Human) 3745, (Mouse) 16500, (Rat) 25736
Molecular Function: voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support