Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: EVKMNHDTE ACSEPSLLST EMLSYQDDEN RQLTLPEEDK RDIRQSPKRG FLRSASLGRR ASFHLECLKR QKDRGGDISQ KTVLPLHLVH HQALAVAGLS PLLQRSHSPA SFPRPFATPP ATPGSRGWPP QPVPTLRLEG VESSEKLNSS FPSIHCGSWA ETTPGGGGSS AARRVRPVSL MVPSQAGAPG RQFHGSASSL VEAVLISEGL GQFAQDPKFI EVTTQELADA CDMTIEEMES AADNILSGGA PQSPNGALLP FVNCRDAGQD RAGGEEDAGC VRARGRPSEE ELQDSRVYVS SL (1835-2135 aa encoded by B C146846)
CACNA1C encodes an alpha-1 subunit of a voltage-dependent calcium channel. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization. The alpha-1 subunit consists of 24 transmembrane segments and forms the pore through which ions pass into the cell. The calcium channel consists of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. There are multiple isoforms of each of these proteins, either encoded by different genes or the result of alternative splicing of transcripts. The protein encoded by CACNA1C binds to and is inhibited by dihydropyridine. Alternative splicing results in many transcript variants encoding different proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain class C; CACH3; CACN4; CACNA 1D; CACNA1C intronic transcript 2; CACNA1C intronic transcript 2 (non-protein coding); CACNL1A2; calcium channel voltage-dependent alpha1c subunit; calcium channel, cardic dihydropyridine-sensitive, alpha-1 subunit; calcium channel, voltage-dependent, alpha 1C subunit; calcium channel, voltage-dependent, L type, alpha 1C subunit; CaV1.2; DHP receptor; DHPR, alpha-1 subunit; ion channel; L-type calcium channel alpha-1 subunit; long-NT; LTCC; MGC120730; neuronal voltage-gated calcium channel alpha 1C subunit; RBC; skeletal muscle-specific calcium channel; unnamed protein product; voltage-dependent calcium channel; voltage-dependent L-type Ca2+ channel alpha 1 subunit; voltage-gated calcium channel subunit alpha Cav1.2
Gene Aliases: CACH2; CACN2; CACNA1C-IT2; CACNL1A1; CaV1.2; CCHL1A1; D930026N18Rik; LQT8; MBC; MELC-CC; NEDHLSS; RATIVS302; TS; TS. LQT8
UniProt ID: (Human) Q13936, (Mouse) Q01815, (Rat) P22002
Entrez Gene ID: (Human) 775, (Mouse) 12288, (Rat) 24239
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support