Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Laminin alpha-5 Antibodies

          Abnova

          LAMA5 Polyclonal Antibody, MaxPab™

          View all (7) Laminin alpha-5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite LAMA5 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          LAMA5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          LAMA5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          LAMA5 Antibody (H00003911-D01P) in WB

          Western Blot analysis of LAMA5 expression in transfected 293T cell line (H00003911-T01) by LAMA5 MaxPab polyclonal antibody.

          Lane 1: LAMA5 transfected lysate(74.00 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          LAMA5 Antibody in Western Blot (WB)
          LAMA5 Antibody in Proximity Ligation Assay (PLA) (PLA)
          LAMA5 Polyclonal Antibody, MaxPab™

          Product Details

          H00003911-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          Proximity Ligation Assay (PLA) (PLA)

          1:1200
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          LAMA5 (AAH03355.1, 1 a.a. approximately 670 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MGVSLRDKKV HWVYQLGEAG PAVLSIDEDI GEQFAAVSLD RTLQFGHMSV TVERQMIQET KGDTVAPGAE GLLNLRPDDF VFYVGGYPST FTPPPLLRFP GYRGCIEMDT LNEEVVSLYN FERTFQLDTA VDRPCARSKS TGDPWLTDGS YLDGTGFARI SFDSQISTTK RFEQELRLVS YSGVLFFLKQ QIQVFLLGGS RKRVLVRVER ATVYSVEQDN DLELADAYYL GGVPPDQLPP SLRRLFPTGG SVRGCVKGIK ALGKYVDLKR LNTTGVSAGC TADLLVGRAM TFHGHGFLRL ALSNVAPLTG NVYSGFGFHS AQDSALLYYR ASPDGLCQVS LQQGRVSLQL LRTEVKTQAG FADGAPHYVA FYSNATGVWL YVDDQLQQMK PHRGPPPELQ PQPEGPPRLL LGGLPESGTI YNFSGCISNV FVQRLLGPQR VFDLQQNLGS VNVSTGCAPA LQAQTPGLGP RGLQATARKA SRRSRQPARH PACMLPPHLR TTRDSYQFGG SLSSHLEFVG ILARHRNWPS LSMHVLPRSS RGLLLFTARL RPGSPSLALF LSNGHFVAQM EGLGTRLRAQ SRQRSRPGRW HKVSVRWEKN RILLVTDGAR AWSQEGPHRQ HQGAEHPQPH TLFVGGLPAS SHSSKLPTSQ ELHCLMWAWN WRCGPWQSPD

          Target Information

          Components of the extracellular matrix exert myriad effects on tissues throughout the body. In particular, the laminins, a family of heterotrimeric extracellular glycoproteins, affect tissue development and integrity in such diverse organs as the kidney, lung, skin, and nervous system. It is thought that laminins mediate the attachment, migration, and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. Laminins function as heterotrimeric complexes of alpha, beta, and gamma chains, with each chain type representing a different subfamily of proteins. The protein encoded by this gene belongs to the alpha subfamily of laminin chains and is a major component of basement membranes. Two transcript variants encoding different isoforms have been found for this gene, but the full-length nature of one of them has not been determined.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: laminin alpha-5 chain; laminin alpha5-chain; Laminin subunit alpha-5; Laminin-10 subunit alpha; Laminin-11 subunit alpha; Laminin-15 subunit alpha; RP11-157P1.6

          View more View less

          Gene Aliases: BBDS2; KIAA0533; KIAA1907; LAMA5; NPHS26

          View more View less

          UniProt ID: (Human) O15230

          View more View less

          Entrez Gene ID: (Human) 3911

          View more View less

          Function(s)
          receptor binding integrin binding extracellular matrix structural constituent receptor agonist activity cell adhesion mediator activity
          Process(es)
          branching involved in ureteric bud morphogenesis morphogenesis of a polarized epithelium neural crest cell migration kidney development hair follicle development cell adhesion integrin-mediated signaling pathway muscle organ development organ morphogenesis morphogenesis of embryonic epithelium cell migration regulation of cell adhesion lung development regulation of cell migration substrate adhesion-dependent cell spreading trunk neural crest cell migration odontogenesis of dentin-containing tooth regulation of embryonic development skeletal system morphogenesis system development branching morphogenesis of an epithelial tube regulation of epithelial cell proliferation cilium morphogenesis branching involved in salivary gland morphogenesis protein localization to plasma membrane postsynapse organization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline