Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: AHNAWQKAM DLDQDVLSAL AEVEQLSKMV SEAKLRADEA KQSAEDILLK TNATKEKMDK SNEELRNLIK QIRNFLTQDS ADLDSIEAVA NEVLKMEMPS TPQQLQNLTE DIRERVESLS QVEVILQHSA ADIARAEMLL EEAKRASKSA TDVKVTADMV KEALEEAEKA QVAAEKAIKQ ADEDIQGTQN LLTSIESETA ASEETLFNAS QRISELERNV EELKRKAAQN SGEAEYIEKV VYTVKQSAED VKKTLDGELD EKYKKVENLI AKKTEESADA RRKAEMLQNE AKTLLAQANS KLQLLKDLER KYEDNQRYLE DKAQELARLE GEVRSLLKDI SQKVAVYSTC L (1437-1786 aa encoded by BC113455)
Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin beta 1; laminin, beta 1; laminin-1 subunit beta; laminin-10 subunit beta; laminin-111; laminin-12 subunit beta; laminin-2 subunit beta; laminin-6 subunit beta; laminin-8 subunit beta; MGC142015; unnamed protein product
Gene Aliases: CLM; D130003D08Rik; Lamb-1; Lamb1-1; LIS5; LKBMH; LUCAO
UniProt ID: (Human) P07942, (Mouse) P02469
Entrez Gene ID: (Human) 3912, (Mouse) 16777, (Rat) 298941
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support