Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Laminin beta-1 Antibodies

          Abnova

          LAMB1 Polyclonal Antibody, MaxPab™

          View all (14) Laminin beta-1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite LAMB1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          LAMB1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          LAMB1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          LAMB1 Antibody (H00003912-B01P) in WB

          Western Blot analysis of LAMB1 expression in transfected 293T cell line by LAMB1 MaxPab polyclonal antibody. Lane 1: LAMB1 transfected lysate(198.1 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          LAMB1 Antibody in Western Blot (WB)

          Product Details

          H00003912-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          LAMB1 (NP_002282.1, 1 a.a. approximately 1786 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MGLLQLLAFS FLALCRARVR AQEPEFSYGC AEGSCYPATG DLLIGRAQKL SVTSTCGLHK PEPYCIVSHL QEDKKCFICN SQDPYHETLN PDSHLIENVV TTFAPNRLKI WWQSENGVEN VTIQLDLEAE FHFTHLIMTF KTFRPAAMLI ERSSDFGKTW GVYRYFAYDC EASFPGISTG PMKKVDDIIC DSRYSDIEPS TEGEVIFRAL DPAFKIEDPY SPRIQNLLKI TNLRIKFVKL HTLGDNLLDS RMEIREKYYY AVYDMVVRGN CFCYGHASEC APVDGFNEEV EGMVHGHCMC RHNTKGLNCE LCMDFYHDLP WRPAEGRNSN ACKKCNCNEH SISCHFDMAV YLATGNVSGG VCDDCQHNTM GRNCEQCKPF YYQHPERDIR DPNFCERCTC DPAGSQNEGI CDSYTDFSTG LIAGQCRCKL NVEGEHCDVC KEGFYDLSSE DPFGCKSCAC NPLGTIPGGN PCDSETGHCY CKRLVTGQHC DQCLPEHWGL SNDLDGCRPC DCDLGGALNN SCFAESGQCS CRPHMIGRQC NEVEPGYYFA TLDHYLYEAE EANLGPGVSI VERQYIQDRI PSWTGAGFVR VPEGAYLEFF IDNIPYSMEY DILIRYEPQL PDHWEKAVIT VQRPGRIPTS SRCGNTIPDD DNQVVSLSPG SRYVVLPRPV CFEKGTNYTV RLELPQYTSS DSDVESPYTL IDSLVLMPYC KSLDIFTVGG SGDGVVTNSA WETFQRYRCL ENSRSVVKTP MTDVCRNIIF SISALLHQTG LACECDPQGS LSSVCDPNGG QCQCRPNVVG RTCNRCAPGT FGFGPSGCKP CECHLQGSVN AFCNPVTGQC HCFQGVYARQ CDRCLPGHWG FPSCQPCQCN GHADDCDPVT GECLNCQDYT MGHNCERCLA GYYGDPIIGS GDHCRPCPCP DGPDSGRQFA RSCYQDPVTL QLACVCDPGY IGSRCDDCAS GYFGNPSEVG GSCQPCQCHN NIDTTDPEAC DKETGRCLKC LYHTEGEHCQ FCRFGYYGDA LRQDCRKCVC NYLGTVQEHC NGSDCQCDKA TGQCLCLPNV IGQNCDRCAP NTWQLASGTG CDPCNCNAAH SFGPSCNEFT GQCQCMPGFG GRTCSECQEL FWGDPDVECR ACDCDPRGIE TPQCDQSTGQ CVCVEGVEGP RCDKCTRGYS GVFPDCTPCH QCFALWDVII AELTNRTHRF LEKAKALKIS GVIGPYRETV DSVERKVSEI KDILAQSPAA EPLKNIGNLF EEAEKLIKDV TEMMAQVEVK LSDTTSQSNS TAKELDSLQT EAESLDNTVK ELAEQLEFIK NSDIRGALDS ITKYFQMSLE AEERVNASTT EPNSTVEQSA LMRDRVEDVM MERESQFKEK QEEQARLLDE LAGKLQSLDL SAAAEMTCGT PPGASCSETE CGGPNCRTDE GERKCGGPGC GGLVTVAHNA WQKAMDLDQD VLSALAEVEQ LSKMVSEAKL RADEAKQSAE DILLKTNATK EKMDKSNEEL RNLIKQIRNF LTQDSADLDS IEAVANEVLK MEMPSTPQQL QNLTEDIRER VESLSQVEVI LQHSAADIAR AEMLLEEAKR ASKSATDVKV TADMVKEALE EAEKAQVAAE KAIKQADEDI QGTQNLLTSI ESETAASEET LFNASQRISE LERNVEELKR KAAQNSGEAE YIEKVVYTVK QSAEDVKKTL DGELDEKYKK VENLIAKKTE ESADARRKAE MLQNEAKTLL AQANSKLQLL KDLERKYEDN QRYLEDKAQE LARLEGEVRS LLKDISQKVA VYSTCL

          Target Information

          Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: laminin B1; Laminin B1 chain; laminin beta 1; Laminin subunit beta-1; laminin, beta 1; Laminin-1 subunit beta; Laminin-10 subunit beta; Laminin-12 subunit beta; Laminin-2 subunit beta; Laminin-6 subunit beta; Laminin-8 subunit beta; MGC142015; unnamed protein product

          View more View less

          Gene Aliases: CLM; LAMB1; LIS5

          View more View less

          UniProt ID: (Human) P07942

          View more View less

          Entrez Gene ID: (Human) 3912

          View more View less

          Function(s)
          integrin binding structural molecule activity extracellular matrix structural constituent protein binding enzyme binding glycosphingolipid binding
          Process(es)
          cell adhesion negative regulation of cell adhesion embryo implantation anatomical structure morphogenesis cell migration neuronal-glial interaction involved in cerebral cortex radial glia guided migration positive regulation of cell migration neuron projection development substrate adhesion-dependent cell spreading endodermal cell differentiation odontogenesis positive regulation of cell adhesion positive regulation of epithelial cell proliferation positive regulation of muscle cell differentiation regulation of basement membrane organization positive regulation of integrin-mediated signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline