Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • LAMP2 (CD107b) Antibodies

          Invitrogen

          LAMP-2A Polyclonal Antibody (AMC2)

          Advanced Verification
          58 Published Figures
          81 References
          View all (69) LAMP2 (CD107b) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite LAMP-2A Polyclonal Antibody (AMC2)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (58)
          • Advanced Verification (1)
          LAMP-2A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          LAMP-2A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 60

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          LAMP-2A Antibody (51-2200) in WB

          Western blot analysis was performed on whole cell extracts (30 µg lysate) of C2C12 (Lane 1), NIH/3T3 (Lane 2) and PC-12 (Lane 3). The blot was probed with Anti-LAMP2 Rabbit Polyclonal Antibody (Product # 51-2200, 2 µg/mL) and detected by chemiluminescence using Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, HRP conjµgate (Product # A27036, 0.4µg/mL, 1:2500 dilution). A ~110 kDa band corresponding to LAMP2 was observed across the cell lines tested. Known quantity of protein samples were electrophoresed using Novex® NuPA... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Immunocytochemistry (ICC/IF)
          LAMP-2A Antibody in Immunocytochemistry (ICC/IF)
          LAMP-2A Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          LAMP-2A Antibody in Immunocytochemistry (ICC/IF)
          LAMP-2A Antibody in Immunocytochemistry (ICC/IF)
          LAMP-2A Antibody in Immunocytochemistry (ICC/IF)
          LAMP-2A Antibody in Immunocytochemistry (ICC/IF)
          LAMP-2A Antibody in Immunocytochemistry (ICC/IF)
          LAMP-2A Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          LAMP-2A Antibody in Immunocytochemistry (ICC/IF)
          LAMP-2A Antibody in Immunocytochemistry (ICC/IF)
          LAMP-2A Antibody in Immunocytochemistry (ICC/IF)
          LAMP-2A Antibody in Immunocytochemistry (ICC/IF)
          LAMP-2A Antibody in Immunocytochemistry (ICC/IF)
          LAMP-2A Antibody in Immunocytochemistry (ICC/IF)
          LAMP-2A Antibody in Immunohistochemistry (IHC)
          LAMP-2A Antibody in Immunohistochemistry (IHC)
          LAMP-2A Antibody in Immunohistochemistry (IHC)
          LAMP-2A Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          LAMP-2A Antibody in Immunohistochemistry (IHC)
          LAMP-2A Antibody in Immunohistochemistry (IHC)
          LAMP-2A Antibody in Immunohistochemistry (IHC)
          LAMP-2A Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody in Western Blot (WB)
          LAMP-2A Antibody
          LAMP-2A Polyclonal Antibody (AMC2)

          Product Details

          51-2200

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-1 µg/mL
          View 46 publications 46 publications

          Immunohistochemistry (IHC)

          -
          View 10 publications 10 publications

          Immunocytochemistry (ICC/IF)

          -
          View 20 publications 20 publications

          ELISA (ELISA)

          0.1-1 µg/mL
          -

          Immunoprecipitation (IP)

          -
          View 2 publications 2 publications

          Dot blot (DB)

          -
          View 1 publication 1 publication

          Inhibition Assays (Inhibition)

          Assay-dependent
          -

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Clone

          AMC2

          Immunogen

          A synthetic peptide of the cytosolic tail of rat LAMP-2A (GLKRHHTGYEQF)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533900

          Product Specific Information

          This antibody was tested for specificity in rat and mouse (it does not cross-react with the human variant) and was demonstrated to be highly selective for LAMP-2A without cross-reacting with LAMP-2B or C. This protein was previously known as lgp96.

          In western blotting this antibody was used at 90 ng/mL with 100 µg of rat liver lysosomal protein. This antibody is also able to completely block transport of cytosolic proteins into lysosomes in rat-derived cells.

          Target Information

          LAMP2 (Lysosome-associated membrane glycoprotein 2, CD107b) is a member of a family of membrane glycoproteins. LAMP2 provides selectins with carbohydrate ligands, and may play a role in tumor cell metastasis, and have a function in the protection, maintenance, and adhesion of the lysosome. Prior to posttranslational modification, LAMP2 is approximately 46 kDa polypeptide. Mature, functional LAMP2 is extensively glycosylated with a variety of different N-linked and O-linked oligosaccharides with a total molecular weight of about 100-130 kDa. LAMP 2 has a structure comprised of a large amino-terminal intra-lysosomal domain, a hydrophobic trans-membrane domain, and a short carboxyl-terminal cytoplasmic tail. LAMP2 is used to monitor a very selective form of autophagy called chaperone-mediated autophagy. LAMP 2 has a structure comprised of a large amino-terminal intra-lysosomal domain, a hydrophobic trans-membrane domain, and a short carboxyl-terminal cytoplasmic tail. The LAMP protein has 3 variant forms: LAMP-2A, -2B, and 2C. LAMP2, together with CD107a/LAMP-1, is a major constituent of lysosomal membrane. The LAMP proteins are involved in lysosome biogenesis and are required for fusion of lysosomes with phagosomes, and LAMP2 is an important regulator in successful phagosomal maturation. LAMP2 deficiency causes an accumulation of autophagosomes in many tissues leading to cardiomyopathy and myopathy (Danon's disease).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CD107 antigen-like family member B; CD107b; Igp96; LAMP-2; LAMP-2 Isoform 2A; LAMP-2 Variant A; LGP-110; LGP-96; LGP-B; lysosomal membrane glycoprotein 2; Lysosomal membrane glycoprotein type B; Lysosome-associated membrane glycoprotein 2; lysosome-associated membrane protein 2; RP23-193O17.2

          View more View less

          Gene Aliases: CD107b; Lamp II; Lamp-2; Lamp-2a; Lamp-2b; Lamp-2c; Lamp2; LGP-B; Mac3

          View more View less

          UniProt ID: (Mouse) P17047, (Rat) P17046

          View more View less

          Entrez Gene ID: (Mouse) 16784, (Rat) 24944

          View more View less

          Function(s)
          enzyme binding protein domain specific binding molecular_function membrane trafficking regulatory protein
          Process(es)
          protein stabilization biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5btrv:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline