Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • LARP7 Antibodies

          Proteintech

          LARP7 Polyclonal Antibody

          13 References
          View all (8) LARP7 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite LARP7 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (20)
          LARP7 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          LARP7 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 20

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          LARP7 Antibody (17067-1-AP) in ICC/IF

          Immunofluorescent analysis of ( 10% Formaldehyde ) fixed HeLa cells using 17067-1-AP (LARP7 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          LARP7 Antibody in Immunocytochemistry (ICC/IF)
          LARP7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LARP7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LARP7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LARP7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LARP7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LARP7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LARP7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LARP7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LARP7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LARP7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LARP7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LARP7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LARP7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LARP7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LARP7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LARP7 Antibody in Western Blot (WB)
          LARP7 Antibody in Western Blot (WB)
          LARP7 Antibody in Western Blot (WB)
          LARP7 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          LARP7 Polyclonal Antibody

          Product Details

          17067-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 9 publications 9 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          LARP7 Fusion Protein Ag10891 (83-331 aa encoded by BC006981)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.13 mg/mL

          Amount

          19.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: LTTDGKLIA RALRSSAVVE LDLEGTRIRR KKPLGERPKD EDERTVYVEL LPKNVNHSWI ERVFGKCGNV VYISIPHYKS TGDPKGFAFV EFETKEQAAK AIEFLNNPPE EAPRKPGIFP KTVKNKPIPA LRVVEEKKKK KKKKGRMKKE DNIQAKEENM DTSNTSISKM KRSRPTSEGS DIESTEPQKQ CSKKKKKRDR VEASSLPEVR TGKRKRSSSE DAESLAPRSK VKKIIQKDII KEASEASKE (83-331 aa encoded by BC006981)

          Target Information

          LARP7 gene ontology annotations related to this gene include RNA processing.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: DKFZP564K112; La ribonucleoprotein domain family member 7; La ribonucleoprotein domain family, member 7; LARP7; P-TEFb-interaction protein for 7SK stability; unnamed protein product

          View more View less

          Gene Aliases: ALAZS; C330027G06Rik; D3Wsu161e; HDCMA18P; hLARP7; PIP7S

          View more View less

          UniProt ID: (Human) Q4G0J3, (Rat) Q5XI01, (Mouse) Q05CL8

          View more View less

          Entrez Gene ID: (Human) 51574, (Rat) 686883, (Mouse) 28036

          View more View less

          Function(s)
          nucleic acid binding RNA binding protein binding U6 snRNA binding 7SK snRNA binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter box C/D snoRNA 3'-end processing RNA processing spermatogenesis negative regulation of viral transcription germ cell proliferation regulation of mRNA splicing, via spliceosome positive regulation of G1/S transition of mitotic cell cycle positive regulation of protein localization to Cajal body positive regulation of snRNA transcription by RNA polymerase II U6 2'-O-snRNA methylation negative regulation of transcription elongation from RNA polymerase II promoter
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-kp6hx:80/100.66.72.150:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0