Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: EFEKSRMGP SGGEGMEPER RDSQDGSSYR RPLEQQQPHH IPTSAPVYQQ PQQQPVAQSY GGYKEPAAPV SIQRSAPGGG GKRYRAA (125-210 aa encoded by BC012460 )
This gene encodes a member of a LIM protein subfamily characterized by a LIM motif and a domain of Src homology region 3. The encoded protein functions as an actin-binding protein and possibly in cytoskeletal organization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LASP-1; Lasp1; metastatic lymph node gene 50 protein; Tg(11Col1a1)NG; unnamed protein product
Gene Aliases: Def-4; LASP-1; MLN 50; MLN50; SH3P6; Tg(Col1a1-lacZ)1Ngma
UniProt ID: (Human) Q14847, (Mouse) Q61792, (Rat) Q99MZ8
Entrez Gene ID: (Human) 3927, (Mouse) 16796, (Rat) 29278
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support