Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • LETM1 Antibodies

          Abnova

          LETM1 Polyclonal Antibody, MaxPab™

          View all (14) LETM1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite LETM1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          LETM1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          LETM1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          LETM1 Antibody (H00003954-D01P) in WB

          LETM1 MaxPab rabbit polyclonal antibody. Western Blot analysis of LETM1 expression in A-431. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          LETM1 Antibody in Western Blot (WB)
          LETM1 Polyclonal Antibody, MaxPab™

          Product Details

          H00003954-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          LETM1 (NP_036450.1, 1 a.a. approximately 739 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MASILLRSCR GRAPARLPPP PRYTVPRGSP GDPAHLSCAS TLGLRNCLNV PFGCCTPIHP VYTSSRGDHL GCWALRPECL RIVSRAPWTS TSVGFVAVGP QCLPVRGWHS SRPVRDDSVV EKSLKSLKDK NKKLEEGGPV YSPPAEVVVK KSLGQRVLDE LKHYYHGFRL LWIDTKIAAR MLWRILNGHS LTRRERRQFL RICADLFRLV PFLVFVVVPF MEFLLPVAVK LFPNMLPSTF ETQSLKEERL KKELRVKLEL AKFLQDTIEE MALKNKAAKG SATKDFSVFF QKIRETGERP SNEEIMRFSK LFEDELTLDN LTRPQLVALC KLLELQSIGT NNFLRFQLTM RLRSIKADDK LIAEEGVDSL NVKELQAACR ARGMRALGVT EDRLRGQLKQ WLDLHLHQEI PTSLLILSRA MYLPDTLSPA DQLKSTLQTL PEIVAKEAQV KVAEVEGEQV DNKAKLEATL QEEAAIQQEH REKELQKRSE VAKDFEPERV VAAPQRPGTE PQPEMPDTVL QSETLKDTAP VLEGLKEEEI TKEEIDILSD ACSKLQEQKK SLTKEKEELE LLKEDVQDYS EDLQEIKKEL SKTGEEKYVE ESKASKRLTK RVQQMIGQID GLISQLEMDQ QAGKLAPANG MPTGENVISV AELINAMKQV KHIPESKLTS LAAALDENKD GKVNIDDLVK VIELVDKEDV HISTSQVAEI VATLEKEEKV EEKEKAKEKA EKEVAEVKS

          Target Information

          This gene encodes a protein that is localized to the inner mitochondrial membrane. The protein functions to maintain the mitochondrial tubular shapes and is required for normal mitochondrial morphology and cellular viability. Mutations in this gene cause Wolf-Hirschhorn syndrome, a complex malformation syndrome caused by the deletion of parts of the distal short arm of chromosome 4. Related pseudogenes have been identified on chromosomes 8, 15 and 19.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Electroneutral mitochondrial K(+)/H(+)exchanger; KHE; LETM1 and EF-hand domain-containing protein 1, mitochondrial; leucine zipper-EF-hand containing transmembrane protein 1; Leucine zipper-EF-hand-containing transmembrane protein 1; Mdm38 homolog; Mitochondrial proton/calcium exchanger protein; unnamed protein product

          View more View less

          Gene Aliases: CONDMIM; KHE; LETM1; Mdm38; SLC55A1

          View more View less

          UniProt ID: (Human) O95202, (Mouse) Q9Z2I0

          View more View less

          Entrez Gene ID: (Human) 3954, (Mouse) 56384

          View more View less

          Function(s)
          calcium ion binding protein binding antiporter activity calcium:proton antiporter activity ribosome binding metal ion binding secondary carrier transporter
          Process(es)
          ion transport potassium ion transport calcium ion transport mitochondrial calcium ion transport mitochondrion organization inner mitochondrial membrane organization protein hexamerization cristae formation protein homooligomerization mitochondrial calcium ion homeostasis negative regulation of mitochondrial calcium ion concentration calcium export from the mitochondrion mitochondrial potassium ion transmembrane transport regulation of cellular hyperosmotic salinity response hydrogen ion transmembrane transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline