Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Recommended positive controls: 293T, H1299.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LGR7 is a low density lipoprotein receptor of 757amino acid belonging to the G-protein coupled receptor 1 family with rhodopsin-like receptor activity. It has seven transmembrane region, cysteine-rich motif, 10 LRR domains and 5 N-glycosylation sites in the N terminus. Research reports show that it has receptor for relaxins (RLN1 and RLN2) which stimulates adenylate cyclase activation, substrate adhesion, and chemo migratory capacity of mononuclear leukocytes. LGR7 is critical for mediating these biological responses by use of RNA interference lentiviral short hairpin constructs. It is widely expressed in 86% of 15 primary EC tissue samples including brain, kidney, testis and endometrium.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: leucine-rich repeat-containing G protein-coupled receptor 7; LGR7; LGR7.1; LGR7.10; LGR7.2; MGC138347; MGC142177; relaxin/insulin like family peptide receptor 1; unnamed protein product
Gene Aliases: LGR7; RXFPR1
UniProt ID: (Human) Q9HBX9
Entrez Gene ID: (Human) 59350
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support