Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • LILRB2 (ILT4) Antibodies

          Proteintech

          LILRB2 Polyclonal Antibody

          View all (26) LILRB2 (ILT4) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite LILRB2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          LILRB2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          LILRB2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          LILRB2 Antibody (14154-1-AP) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed THP-1 cells using LILRB2 antibody (14154-1-AP) at dilution of 1:200 and CoraLite®488-Conjugated Goat Anti-Rabbit IgG (H+L) (SA00013-2). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          LILRB2 Antibody in Immunocytochemistry (ICC/IF)
          LILRB2 Antibody in Western Blot (WB)
          LILRB2 Polyclonal Antibody

          Product Details

          14154-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          LILRB2 fusion protein Ag5336
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          75 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: GTIPKPTLWA EPDSVITQGS PVTLSCQGSL EAQEYRLYRE KKSASWITRI RPELVKNGQF HIPSITWEHT GRYGCQYYSR ARWSELSDPL VLVMTGAYPK PTLSAQPSPV VTSGGRVTLQ CESQVAFGGF ILCKEGEDEH PQCLNSQPHA RGSSRAIFSV GPVSPNRRWS HRCYGYDLNS PYVWSSPSDL LELLVPGVSK KPSLSVQPGP VVAPGESLTL QCVSDVGYDR FVLYKEGERD LRQLPGRQPQ AGLSQANFTL GPVSRSYGGQ YRCYGAYNLS SEWSAPSDPL DILITGQIHG TPFISVQPGP TVASGENVTL LCQSWRQ

          Target Information

          CD85d, also known as LILRB2 or ILT4, is a member of the immunoglobulin-like transcript (ILT), leukocyte Ig-like receptor (LIR), and monocyte Ig-like receptor (MIR) family. It is a 110 kDa single transmembrane receptor with three Ig-like domains and a long cytoplasmic domain containing immunoreceptor tyrosine-based inhibitory motifs (ITIMs). CD85d is expressed on monocytes, macrophages, and dendritic cells. The receptor binds to non-classical class I molecules such as HLA-G, -B, and G1. Upon ligand binding, the tyrosine phosphatase SHP-1 interacts with the ITIMs, transducing a negative signal that inhibits immune response stimulation. This interaction is thought to contribute to tolerance during fetal implantation and plays a role in controlling inflammatory responses and cytotoxicity, helping to focus the immune response and limit autoreactivity. CD85d is also involved in organ allograft rejection, with the levels of ILT3 and ILT4 on dendritic cells suggesting its role in this process. The LILRB2 gene, located in a gene cluster at chromosomal region 19q13.4, encodes multiple transcript variants with different isoforms, highlighting its complexity and importance in immune regulation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CD85 antigen-like family member D; CD85d; CD85d antigen; Ig-like transcript 4; ILT-4; immunoglobulin superfamily; Immunoglobulin-like transcript 4; leucocyte Ig-like receptor B2; Leukocyte immunoglobulin-like receptor subfamily B member 2; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 2; LIR-2; LIR2CD85D; MIR-10; MIR10LILRA6; monocyte/macrophage Ig-related receptor; Monocyte/macrophage immunoglobulin-like receptor 10; myeloid inhibitory receptor 10; unnamed protein product

          View more View less

          Gene Aliases: CD85D; ILT-4; ILT4; LILRB2; LIR-2; LIR2; MIR-10; MIR10

          View more View less

          UniProt ID: (Human) Q8N423

          View more View less

          Entrez Gene ID: (Human) 10288

          View more View less

          Function(s)
          beta-amyloid binding protein binding protein phosphatase 1 binding MHC class Ib protein complex binding MHC class Ib protein binding inhibitory MHC class I receptor activity MHC class I protein binding protein homodimerization activity macromolecular complex binding cell adhesion molecule binding
          Process(es)
          adaptive immune response immune system process negative regulation of antigen processing and presentation positive regulation of tolerance induction positive regulation of T cell tolerance induction immune response-regulating signaling pathway immune response-inhibiting cell surface receptor signaling pathway Fc receptor mediated inhibitory signaling pathway immune response cellular defense response signal transduction cell surface receptor signaling pathway cell-cell signaling learning or memory positive regulation of interleukin-6 production heterotypic cell-cell adhesion positive regulation of T cell proliferation negative regulation of T cell proliferation positive regulation of regulatory T cell differentiation negative regulation of protein metabolic process negative regulation of calcium ion transport cellular response to lipopolysaccharide interleukin-10-mediated signaling pathway regulation of long-term synaptic potentiation positive regulation of long term synaptic depression negative regulation of postsynaptic density organization negative regulation of T cell costimulation regulation of dendritic cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline