Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • LLGL2 Antibodies

          Abnova

          LLGL2 Polyclonal Antibody, MaxPab™

          View all (8) LLGL2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite LLGL2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          LLGL2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          LLGL2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          LLGL2 Antibody (H00003993-B01P) in WB

          Western Blot analysis of LLGL2 expression in transfected 293T cell line by LLGL2 MaxPab polyclonal antibody. Lane 1: LLGL2 transfected lysate(112.2 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          LLGL2 Antibody in Western Blot (WB)

          Product Details

          H00003993-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          LLGL2 (AAH64994.1, 1 a.a. approximately 1020 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MRRFLRPGHD PVRERLKRDL FQFNKTVEHG FPHQPSALGY SPSLRILAIG TRSGAIKLYG APGVEFMGLH QENNAVTQIH LLPGQCQLVT LLDDNSLHLW SLKVKGGASE LQEDESFTLR GPPGAAPSAT QITVVLPHSS CELLYLGTES GNVFVVQLPA FRALEDRTIS SDAVLQRLPE EARHRRVFEM VEALQEHPRD PNQILIGYSR GLVVIWDLQG SRVLYHFLSS QQLENIWWQR DGRLLVSCHS DGSYCQWPVS SEAQQPEPLR SLVPYGPFPC KAITRILWLT TRQGLPFTIF QGGMPRASYG DRHCISVIHD GQQTAFDFTS RVIGFTVLTE ADPAATFDDP YALVVLAEEE LVVIDLQTAG WPPVQLPYLA SLHCSAITCS HHVSNIPLKL WERIIAAGSR QNAHFSTMEW PIDGGTSLTP APPQRDLLLT GHEDGTVRFW DASGVCLRLL YKLSTVRVFL TDTDPNENLS AQGEDEWPPL RKVGSFDPYS DDPRLGIQKI FLCKYSGYLA VAGTAGQVLV LELNDEAAEQ AVEQVEADLL QDQEGYRWKG HERLAARSGP VRFEPGFQPF VLVQCQPPAV VTSLALHSEW RLVAFGTSHG FGLFDHQQRR QVFVKCTLHP SDQLALEGPL SRVKSLKKSL RQSFRRMRRS RVSSRKRHPA GPPGEAQEGS AKAERPGLQN MELAPVQRKI EARSAEDSFT GFVRTLYFAD TYLKDSSRHC PSLWAGTNGG TIYAFSLRVP PAERRMDESV RAEQAKEIQL MHRAPVVGIL VLDGHSVPLP EPLEVAHDLS KSPDMQGSHQ LLVVSEEQFK VFTLPKVSAK LKLKLTALEG SRVRRVSVAH FGSRRAEDYG EHHLAVLTNL GDIQVVSLPL LKPQVRYSCI RREDVSGIAS CVFTKYGQGF YLISPSEFER FSLSTKWLVE PRCLVDSAET KNHRPGNGAG PKKAPSRARN SGTQSDGEEK QPGLVMERAL LSDERVLKEI QSTLEGDRGS GNWRSHRAAV GCSLSNGGAE

          Target Information

          LLGL1 is a protein that is similar to a tumor suppressor in Drosophila. The protein is part of a cytoskeletal network and is associated with nonmuscle myosin II heavy chain and a kinase that specifically phosphorylates this protein at serine residues. The gene for LLGL1 is located within the Smith-Magenis syndrome region on chromosome 17. LLGL2 is a protein similar to lethal (2) giant larvae of Drosophila. In fly, the protein's ability to localize cell fate determinants is regulated by the atypical protein kinase C (aPKC). In human, this protein interacts with aPKC-containing complexes and is cortically localized in mitotic cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: HGL; human giant larvae homolog; lethal giant larvae homolog 2, scribble cell polarity complex component; Lethal(2) giant larvae protein homolog 2; LLGL scribble cell polarity complex component 2; LLGL2, scribble cell polarity complex component; unnamed protein product

          View more View less

          Gene Aliases: HGL; Hugl-2; LGL2; LLGL2

          View more View less

          UniProt ID: (Human) Q6P1M3

          View more View less

          Entrez Gene ID: (Human) 3993

          View more View less

          Function(s)
          GTPase activator activity protein binding PDZ domain binding myosin II binding
          Process(es)
          placenta development exocytosis Golgi to plasma membrane transport regulation of Notch signaling pathway post-embryonic development leucine transport establishment or maintenance of polarity of embryonic epithelium cortical actin cytoskeleton organization regulation of establishment or maintenance of cell polarity multicellular organism growth establishment or maintenance of epithelial cell apical/basal polarity establishment of spindle orientation cell division branching involved in labyrinthine layer morphogenesis labyrinthine layer blood vessel development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0