Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: GDPAVALPH RRFEYKYSFK GPHLVQSDGT VPFWAHAGNA IPSSDQIRVA PSLKSQRGSV WTKTKAAFEN WEVEVTFRVT GRGRIGADGL AIWYAENQGL EGPVFGSADL WNGVGIFFDS FDNDGKKNNP AIVIIGNNGQ IHYDHQNDGA SQALASCQRD FRNKPYPVRA KITYYQNTLT VMINNGFTPD KNDYEFCAKV ENMIIPAQGH FGISAATGGL ADDHDVLSFL TFQLTEPGKE PPTPDKEISE KEKEKYQEEF EHFQQELDKK KEEFQKGHPD LQGQPAEEIF ESVGDRELRQ VFEGQNRIHL EIKQLNRQLD MILDEQRRYV SSLTEEISKR GAGMPGQHGQ ITQQELDTVV KTQHEILR (35-401 aa encoded by BC032330 )
The protein encoded by this gene is a type I integral membrane protein localized in the intermediate region between the endoplasmic reticulum and the Golgi, presumably recycling between the two compartments. The protein is a mannose-specific lectin and is a member of a novel family of plant lectin homologs in the secretory pathway of animal cells. Mutations in the gene are associated with a coagulation defect. Using positional cloning, the gene was identified as the disease gene leading to combined factor V-factor VIII deficiency, a rare, autosomal recessive disorder in which both coagulation factors V and VIII are diminished.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: endoplasmic reticulum-golgi intermediate compartment protein 53; ER-Golgi intermediate compartment 53 kDa protein; ERGIC53; intracellular mannose-specific lectin MR60; unnamed protein product
Gene Aliases: 2610020P13Rik; C730041J05; ERGIC-53; ERGIC53; F5F8D; FMFD1; gp58; MCFD1; MR60; P58
UniProt ID: (Human) P49257, (Rat) Q62902, (Mouse) Q9D0F3
Entrez Gene ID: (Human) 3998, (Rat) 116666, (Mouse) 70361
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support