Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • LMBR1L Antibodies

          Proteintech

          LMBR1L Polyclonal Antibody

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite LMBR1L Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          LMBR1L Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          LMBR1L Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          LMBR1L Antibody (12154-1-AP) in WB

          Mouse testis tissue were subjected to SDS PAGE followed by western blot with 12154-1-AP (LMBR1L antibody) at dilution of 1:600 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          LMBR1L Antibody in Western Blot (WB)
          LMBR1L Antibody in Western Blot (WB)
          LMBR1L Antibody in Western Blot (WB)
          LMBR1L Antibody in Western Blot (WB)
          LMBR1L Antibody in Western Blot (WB)
          LMBR1L Antibody in Flow Cytometry (Flow)
          LMBR1L Antibody in Immunoprecipitation (IP)
          LMBR1L Polyclonal Antibody

          Product Details

          12154-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cells
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          LMBR1L Fusion Protein Ag2787 (219-490 aa encoded by BC015015)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.13 mg/mL

          Amount

          19.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: RMFSVTGKL LVKPRLLEDL EEQLYCSAFE EAALTRRICN PTSCWLPLDM ELLHRQVLAL QTQRVLLEKR RKASAWQRNL GYPLAMLCLL VLTGLSVLIV AIHILELLID EAAMPRGMQG TSLGQVSFSK LGSFGAVIQV VLIFYLMVSS VVGFYSSPLF RSLRPRWHDT AMTQIIGNCV CLLVLSSALP VFSRTLGLTR FDLLGDFGRF NWLGNFYIVF LYNAAFAGLT TLCLVKTFTA AVRAELIRAF GLDRLPLPVS GFPQASRKTQ HQ (219-490 aa encoded by BC015015)

          Target Information

          LMBR1L, also known as LIMR (lipocalin-1-interacting membrane receptor), interacts with lipocalin-1 (Lcn-1), a lipocalin member produced by a number of secretory glands and tissues and known to bind a variety of lipophilic compounds. LMBR1L is a 489-amino acid protein with a predicted molecular mass of 55 kDa and 9 putative transmembrane helices. LMBR1L has been shown to mediate cellular uptake of LMBR1L, thus it is characterized as a endocytic receptor. Alternatively spliced transcript variants encoding different isoforms have been described.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: limb development membrane protein 1 like-protein; limb region 1 homolog (mouse)-like isoform LMBR1L_1; limb region 1 homolog (mouse)-like isoform LMBR1L_2; limb region 1 homolog (mouse)-like isoform LMBR1L_3; limb region 1 homolog (mouse)-like isoform LMBR1L_4; limb region 1 homolog (mouse)-like isoform LMBR1L_5; limb region 1 homolog-like; limb region 1 like; Limb region 1 protein homolog-like; limb region 1-like homolog; limb region 1-like protein -like; LIMR; lipocalin interacting membrane receptor; lipocalin-1 interacting membrane receptor; Lipocalin-1-interacting membrane receptor; lipocalin-interacting membrane receptor; Protein LMBR1L; putative membrane protein; unnamed protein product; Uteroglobin receptor

          View more View less

          Gene Aliases: 1110013E13Rik; AA415653; D15Ertd735e; KIAA1174; LIMR; LMBR1L; RGD1309631; UNQ458/PRO783

          View more View less

          UniProt ID: (Human) Q6UX01, (Mouse) Q9D1E5

          View more View less

          Entrez Gene ID: (Human) 55716, (Rat) 300215, (Mouse) 74775

          View more View less

          Function(s)
          transmembrane signaling receptor activity protein binding molecular_function transmembrane signal receptor
          Process(es)
          endocytosis receptor-mediated endocytosis signal transduction Wnt signaling pathway T cell differentiation T cell proliferation hematopoietic stem cell differentiation T cell apoptotic process negative regulation of canonical Wnt signaling pathway biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline