Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MEELEKELD VVDPDGSVPV GLRQRNQTEK QSTGVYNREA MLNFCEKETK KLMQREMSMD ESKQVETKTD AKNGEERGRD ASKKALGPRR DSDLGKEPKR GGLKKSFSRD RDEAGGKSGE KPKEEKIIRG IDKGRVRAAV DKKEAGKDGR GEERAVATKK EEEKKGSDRN TGLSRDKDKK REEMKEVAKK EDDEKVKGGA PAAPPPPPPP LAPPLIMENL KNSLSPATQR KMGDKVLPAQ EKNSRDQLLA AIRSSNLKQL KKVEVPKLLQ (1-270 aa encoded by BC001755)
The leiomodin 1 protein (LMOD1) has a putative membrane-spanning region and 2 types of tandemly repeated blocks. The transcript is expressed in all tissues tested, with the highest levels in thyroid, eye muscle, skeletal muscle, and ovary. Increased expression of leiomodin 1 may be linked to Graves' disease and thyroid-associated ophthalmopathy. The leiomodin 1 protein has a putative membrane-spanning region and 2 types of tandemly repeated blocks. The transcript is expressed in all tissues tested, with the highest levels in thyroid, eye muscle, skeletal muscle, and ovary. Increased expression of leiomodin 1 may be linked to Graves' disease and thyroid-associated ophthalmopathy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 64 kDa autoantigen 1D; 64 kDa autoantigen 1D3; 64 kDa autoantigen D1; leimodin 1 (smooth muscle); leiomodin 1 (smooth muscle); leiomodin, muscle form; SM-LMOD; smooth muscle leiomodin; thyroid and eye muscle autoantigen D1 (64kD); thyroid-associated ophthalmopathy autoantigen; unnamed protein product
Gene Aliases: 1D; 64kD; 9530015K06Rik; D1; MMIHS3; SM-LMOD; SMLMOD
UniProt ID: (Human) P29536, (Mouse) Q8BVA4, (Rat) A0A0G2K0D3
Entrez Gene ID: (Human) 25802, (Mouse) 93689, (Rat) 304816
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support