Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: CSQGRCENT EGSFLCICPA GFMASEEGTN CIDVDECLRP DVCGEGHCVN TVGAFRCEYC DSGYRMTQRG RCEDIDECLN PSTCPDEQCV NSPGSYQCVP CTEGFRGWNG QCLDVDECLE PNVCANGDCS NLEGSYMCSC H (600-739 aa encoded by BC130289)
The protein encoded by this gene belongs to the family of latent TGF-beta binding proteins. The secretion and activation of TGF-betas is regulated by their association with latency-associated proteins and with latent TGF-beta binding proteins. The product of this gene targets latent complexes of transforming growth factor beta to the extracellular matrix, where the latent cytokine is subsequently activated by several different mechanisms. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LTBP-1; MGC163161; TGF-beta1-BP-1; transforming growth factor beta-1-binding protein 1; transforming growth factor-beta 1 binding protein precursor; unnamed protein product
Gene Aliases: ARCL2E
UniProt ID: (Human) Q14766
Entrez Gene ID: (Human) 4052
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support