PA1-30176 detects LTBR from human samples.
PA1-30176 has been successfully used in flow cytometry, Western blot applications.
The PA1-30176 immunogen is: Recombinant human lymphotoxin betaR extracellular domain, expressed in mouse NSO cells.
The protein encoded by this gene is a member of the tumor necrosis factor (TNF) family of receptors. It is expressed on the surface of most cell types, including cells of epithelial and myeloid lineages, but not on T and B lymphocytes. The protein specifically binds the lymphotoxin membrane form (a complex of lymphotoxin-alpha and lymphtoxin-beta). The encoded protein and its ligand play a role in the development and organization of lymphoid tissue and tranformed cells. Activation of the encoded protein can trigger apoptosis.