Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
11714-1-AP can detect the protein and its dimer and tetramer.
Immunogen sequence: VPVNWEPPQ PLPFPKYLRC YRCLLETKEL GCLLGSDICL TPAGSSCITL HKKNSSGSDV MVSDCRSKEQ MSDCSNTRTS PVSGFWIFSQ YCFLDFCNDP QNRGLYTP (43-150 aa encoded by BC141637 )
LY6G5C belongs to a cluster of leukocyte antigen-6 (LY6) genes located in the major histocompatibility complex (MHC) class III region on chromosome 6. Members of the LY6 superfamily typically contain 70 to 80 amino acids, including 8 to 10 cysteines. Most LY6 proteins are attached to the cell surface by a glycosylphosphatidylinositol (GPI) anchor that is directly involved in signal transduction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lymphocyte antigen 6 complex locus protein G5c; lymphocyte antigen 6 complex, locus G5C; lymphocyte antigen-6 G5C
Gene Aliases: C6orf20; G5C; LY6G5C; LY6G5CA; LY6G5CB; NG33
UniProt ID: (Human) Q5SRR4, (Mouse) Q8K1T5
Entrez Gene ID: (Human) 80741, (Mouse) 114652
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support