Search
Search
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 5
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
LIN-28 is a highly conserved, RNA-binding, cytoplasmic protein. It consists of a cold shock domain and retroviral-type (CCHC) zinc finger motifs that were first identified in Caenorhabditis elegans. LIN-28 controls the timing of events during embryonic development and is readily expressed in embryos, embryonic stem cells and embryonal carcinoma cells. The presence of LIN-28 persists in some adult tissues including cardiac and skeletal muscle. In differentiating myoblasts, LIN-28 increases protein synthesis efficiency and binds to the growth and differentiation factor IGF-II.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CSD-containing, CCHC zinc finger motif containing; CSD-containing; CCHC zinc finger motif-containing protein; lin-28 homolog A; lin-28 homolog B; Lin-28.2; lin-28B; protein lin-28 homolog A-like; RNA-binding protein LIN-28; testis expressed gene 17; testis-expressed protein 17; unnamed protein product; zinc finger CCHC domain-containing protein 1; zinc finger, CCHC domain containing 1
Gene Aliases: 2810403D23Rik; CSDD1; CSDD2; D030047M17Rik; Gm10299; LIN-28; Lin-28.2; lin-28A; LIN28; RGD1566408; Tex17; ZCCHC1
UniProt ID: (Human) Q9H9Z2, (Human) Q6ZN17, (Mouse) Q8K3Y3, (Mouse) Q45KJ6
Entrez Gene ID: (Human) 79727, (Human) 389421, (Mouse) 83557, (Rat) 500562, (Rat) 689054, (Mouse) 380669
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support