MA1-10752 detects Lidocaine.
MA1-10752 has been successfully used in ELISA applications.
The MA1-10752 immunogen is Lidocaine, conjugated to KLH.
Lidocaine is a synthetic aminoethylamide with local anesthetic and antiarrhythmic properties. Lidocaine stabilizes the neuronal membrane by binding to and inhibiting voltage-gated sodium channels, thereby inhibiting the ionic fluxes required for the initiation and conduction of impulses and effecting local anesthesia. [National Center for Biotechnology Information. PubChem Compound Database; CID=3676].
Protein Aliases: Anestacon; C14H22N2O; Lidoderm; Lignocaine; Xylocaine