MA1-10790 detects Mycobacterim avium.
MA1-10790 has been successfully used in ELISA applications.
The MA1-10790 immunogen is M. avium cell extract.
Mycobacteria are straight or slightly curved, non motile rods that cause TB. Mycobacteria avium causes TB in birds and a pulmonary infection of humans. It can also be transmitted to immunocompromised humans such as AIDS patients where it can cause disseminated Mycobacterium avium complex (MAC).
Protein Aliases: m avium; M. avium