MA1-10796 detects E. coli K99 pili. These antibodies do not cross-react with k99-negative E.Coli, E.Coli K88ac, E.Coli 987P, Proteus vulgaris, Pseudomonas aeruginosa, Enterbacter aerogenes, Citrobacter freundii, Klebsiella pneumoniae, or strep. agalactiae.
MA1-10796 has been successfully used in ELISA applications.
The MA1-10796 immunogen is purified E. coli K99 pili.
Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.
Protein Aliases: Escherichia coli