    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO and CRO Solutions
          • Primary Antibodies ›
          • Myc Tag Antibodies
          Invitrogen
          Myc Tag Monoclonal Antibody (Myc.A7), DyLight™ 800 4X PEG
          3 Published Figures
          4 References
          View all (44) Myc Tag antibodies
          Tech Support
          Cite Myc Tag Monoclonal Antibody (Myc.A7), DyLight™ 800 4X PEG
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          • Published Figures (3)
          Myc Tag Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Myc Tag Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          Myc Tag Antibody (MA1-21316-D800) in WB
          Western blot analysis of Myc Epitope Tag was performed by loading 2 µg of E. coli lysate containing a Myc-epitope tagged protein onto a 4-20% Tris-HCl polyacrylamide gel. Proteins were transferred to a low fluorescence PVDF membrane (Product # 22860) and blocked with SEA BLOCK (Product # 37527) for at least 1 hour. The membrane was probed with a DyLight 800-conjugated Myc Epitope Tag monoclonal antibody (Product # MA1-21316-D800) at a dilution of 1:500 overnight at 4°C on a rocking platform and washed in TBS-0.1% Tween-20. Fluorescent dete... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Myc Tag Antibody in Western Blot (WB)
          Myc Tag Antibody in Western Blot (WB)
          Myc Tag Antibody in Western Blot (WB)
          Myc Tag Antibody in Western Blot (WB)
          Product Details
          MA1-21316-D800
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          View 3 publications 3 publications
          Miscellaneous PubMed (Misc)
          -
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Tag
          Published species
          Tag
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          Myc.A7
          Immunogen
          EQKLISEEDL synthetic peptide conjugated to KLH
          Conjugate
          DyLight™ 800 4X PEG DyLight™ 800 4X PEG DyLight™ 800 4X PEG
          • Unconjugated
          • Biotin 
          • DyLight 488
          • DyLight 550
          • DyLight 650
          • DyLight 680
          • HRP
          • Request custom conjugation
          Excitation/Emission Max
          789/794 nm View spectra spectra
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          50 µg
          Purification
          Protein A
          Storage buffer
          MES with proprietary stabilizer
          Contains
          no preservative
          Storage conditions
          4°C, do not freeze
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2536996
          Product Specific Information
          MA1-21316-D800 detects Myc tag from various samples. MA1-21316-D800 has been successfully used for fluorescent detection of Myc tagged proteins in Western Blotting.
          DyLight 800 4X PEG has an excitation/emission of 783/797 nm.
          Target Information
          Myc is a proto-oncogene and encodes a nuclear phosphoprotein that plays a role in cell cycle progression, apoptosis and cellular transformation. The encoded protein forms a heterodimer with the related transcription factor MAX. This complex binds to the E box DNA consensus sequence and regulates the transcription of specific target genes. Amplification of this gene is frequently observed in numerous human cancers. Translocations involving this gene are associated with Burkitt lymphoma and multiple myeloma in human patients. There is evidence to show that translation initiates both from an upstream, in-frame non-AUG (CUG) and a downstream AUG start site, resulting in the production of two isoforms with distinct N-termini.
          Bioinformatics
          Protein Aliases: 46-0603; c Myc; c-myc epitope tag; cmyc; Myc protein; Myc Tag; R950-25; R95025
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline