    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • Complement Factor I Antibodies
          Invitrogen
          Complement Factor I Monoclonal Antibody (OX-21)
          View all (16) Complement Factor I antibodies
          Tech Support
          Cite Complement Factor I Monoclonal Antibody (OX-21)
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          Product Details
          MA1-70056
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Flow Cytometry (Flow)
          1 µg/1 million cells
          -
          ELISA (ELISA)
          1:40,000
          -
          Immunoprecipitation (IP)
          Assay-dependent
          -
          Radioimmune Assays (RIA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          OX-21
          Immunogen
          Human complement Factor I.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • Biotin 
          • FITC
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          250 µg
          Purification
          Protein G
          Storage buffer
          PBS
          Contains
          0.02% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_1073853
          Product Specific Information
          MA1-70056 detects Complement Factor I in human samples.
          MA1-70056 has been successfully used in flow cytometry, ELISA, RIA, immunoprecipitation and Western blot procedures.
          The MA1-70056 immunogen is human complement Factor I.
          Target Information
          Factor I, a serine protease, is a regulatory component of both the alternative and the classical complement pathway. Factor I is synthesized as a single chain precursor of 88 kDa, processed to yield the active form, composed of two disulfide-linked chains, the a-chain (50 kDa) and the b-chain (38 kDa). Plasma concentration is 35 µg/mL.
          Bioinformatics
          Protein Aliases: C3b-inactivator; C3B/C4B inactivator; CFI; complement component I; complement control protein factor I; complement factor I heavy chain; factor i; Konglutinogen-activating factor; light chain of factor I; prepro-C3b/C4B inactivator; unnamed protein product
          View more View less
          Gene Aliases: AHUS3; ARMD13; C3b-INA; C3BINA; FI; IF; KAF
          View more View less
          UniProt ID: (Human) P05156
          View more View less
          Entrez Gene ID: (Human) 3426
          View more View less
          Function(s)
          serine-type endopeptidase activity protein binding serine-type peptidase activity
          Process(es)
          proteolysis complement activation protein processing regulation of complement activation, alternative pathway
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-blue-74d7b4b4d9-vlj45:80/100.66.77.38:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline