    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Cytomegalovirus pp65 Antibodies
          Invitrogen
          Cytomegalovirus pp65 Monoclonal Antibody (I1010D), FITC
          3 Published Figures
          4 References
          View all (4) Cytomegalovirus pp65 antibodies
          Tech Support
          Cite Cytomegalovirus pp65 Monoclonal Antibody (I1010D), FITC
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Published Figures (3)
          Cytomegalovirus pp65 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          Cytomegalovirus pp65 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          Cytomegalovirus pp65 Antibody (MA1-7296) in Flow
          Oncoimmunology 2017 - Figure 1. HCMV positivity in GBM determined comparatively by serology, qPCR and IHC. GBM patients were investigated for presence of HCMV pp65 and IE-1 expression by (A) qPCR, IHC and serology (dark gray bars = positive samples and light gray bar = negative samples) and (B) IE-1 expression on macrophage/myeloid derived cells (left panel) and on tumor cells (right panel). Magnification 400X, Scale bar 100 mum... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Cytomegalovirus pp65 Antibody in Flow Cytometry (Flow)
          Cytomegalovirus pp65 Antibody in Western Blot (WB)
          Cytomegalovirus pp65 Antibody in Western Blot (WB)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          MA1-7296
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5-1:20
          View 2 publications 2 publications
          Immunocytochemistry (ICC/IF)
          1:5-1:20
          -
          Flow Cytometry (Flow)
          -
          View 1 publication 1 publication
          Miscellaneous PubMed (Misc)
          -
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Virus
          Published species
          Virus
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          I1010D
          Immunogen
          Purified CMV pp65.
          Conjugate
          FITC FITC FITC
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          498/517 nm View spectra spectra
          Form
          Liquid
          Concentration
          100 µg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.2, with 10mg/mL BSA
          Contains
          0.1% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_1016993
          Product Specific Information
          The MA1-7296 antibody reacts with Cytomegalovirus pp65 in viral and infected tissue samples.
          MA1-7296 has been successfully used in Western blot and Immunofluorescence applications. Acetone fixation of the antigen source is recommended prior to staining.
          The MA1-7296 antibody was raised against purified CMV pp65.
          Target Information
          Cytomegalovirus (CMV) is an opportunistic pathogen infecting lung, kidney, gut and other organs in situations where an individual is immunologically immature, such as the foetus and neonate. Infection also occurs in immunosuppressed patients e.g. transplant patients, patients undergoing chemotherapy and those with HIV infection. Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5).
          Bioinformatics
          Protein Aliases: CMV; Cytomegalovirus
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-6b866bbc8c-vkppw:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0