    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Parainfluenza Virus Type 1 Antibodies
          Invitrogen
          Parainfluenza Virus Type 1 Monoclonal Antibody (E14D), FITC
          View all (7) Parainfluenza Virus Type 1 antibodies
          Tech Support
          Cite Parainfluenza Virus Type 1 Monoclonal Antibody (E14D), FITC
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          Product Details
          MA1-7314
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:5-1:20
          -
          Product Specifications
          Species Reactivity
          Virus
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          E14D
          Immunogen
          Parainfluenza Type 1 antigen.
          Conjugate
          FITC FITC FITC
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          498/517 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.2, with 10mg/mL BSA
          Contains
          0.1% sodium azide
          Storage conditions
          4°C, store in dark
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_1018115
          Product Specific Information
          The MA1-7314 antibody reacts with Parainfluenza virus 1 in viral and infected tissue samples.
          MA1-7314 has been successfully used in Immunofluorescence applications.
          The MA1-7314 antibody was raised against Parainfluenza Type 1 antigen.
          Bioinformatics
          Protein Aliases: canine parainfluenza virus; hPIV-1; PIV-1
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-74d7b4b4d9-p95z8:80/100.66.74.151:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline