    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • Hepatitis C Virus NS3 Antibodies
          Invitrogen
          Hepatitis C Virus NS3 Monoclonal Antibody (C44D)
          1 Reference
          View all (13) Hepatitis C Virus NS3 antibodies
          Tech Support
          Cite Hepatitis C Virus NS3 Monoclonal Antibody (C44D)
          Additional Information:
          {{banner.description}}
          Product Details
          MA1-7345
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:10-1:50
          -
          Immunocytochemistry (ICC/IF)
          1:10-1:50
          -
          ELISA (ELISA)
          1:20-1:200
          -
          Miscellaneous PubMed (Misc)
          -
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Virus
          Published species
          Virus
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          C44D
          Immunogen
          NS3 protein.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.2
          Contains
          0.1% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_1017476
          Product Specific Information
          The MA1-7345 antibody reacts with Hepatitis C virus NS3 in viral and infected tissue samples.
          MA1-7345 has been successfully used in ELISA, Immunofluorescence and Western blot applications.
          The MA1-7345 antibody was raised against NS3 protein.
          Target Information
          Hepatitis C virus (HCV) is a small (55-65 nm in size), enveloped, positive-sense single-stranded RNA virus of the family Flaviviridae. The hepatitis C virus is the cause of hepatitis C and some cancers such as liver cancer (hepatocellular carcinoma, abbreviated HCC) and lymphomas in humans. Nonstructural protein 3 (NS3), also known as p-70, is a viral nonstructural protein that is 70 kDa cleavage product of the hepatitis C virus polyprotein. It acts as a serine protease. C-terminal two thirds of the protein also acts as helicase and nucleoside triphosphatase. First (N-terminal) 180 aminoacids of NS3 has additional role as cofactor domains for NS2 protein.
          Bioinformatics
          Protein Aliases: HCV; HCV NS3; Hepatitis C Virus; HepC; HepC NS3
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline