    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • gamma Tubulin Antibodies
          Invitrogen
          gamma Tubulin Monoclonal Antibody (4D11)
          Advanced Verification
          8 Published Figures
          30 References
          View all (24) gamma Tubulin antibodies
          Tech Support
          Cite gamma Tubulin Monoclonal Antibody (4D11)
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (6)
          • Published Figures (8)
          • Advanced Verification (1)
          gamma Tubulin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          gamma Tubulin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 15
          gamma Tubulin Antibody (MA1-850) in ICC/IF
          Immunofluorescent analysis of g-Tubulin using anti-g-Tubulin monoclonal antibody (Product # MA1-850, Clone 4D11) (shown in green) in HeLa (A) and U2-OS cells (B). Formalin fixed cells were permeabilized with 0.1% Triton X-100 in TBS for 10 minutes at room temperature. Cells were then blocked with 1% Blocker BSA (Product # 37525) for 15 minutes at room temperature. Cells were probed with a mouse monoclonal a... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          gamma Tubulin Antibody in Immunocytochemistry (ICC/IF)
          gamma Tubulin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          gamma Tubulin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          gamma Tubulin Antibody in Western Blot (WB)
          gamma Tubulin Antibody in Western Blot (WB)
          gamma Tubulin Antibody in Western Blot (WB)
          gamma Tubulin Antibody in Western Blot (WB)
          gamma Tubulin Antibody in Immunocytochemistry (ICC/IF)
          gamma Tubulin Antibody in Western Blot (WB)
          gamma Tubulin Antibody in Western Blot (WB)
          gamma Tubulin Antibody in Western Blot (WB)
          gamma Tubulin Antibody in Western Blot (WB)
          gamma Tubulin Antibody in Western Blot (WB)
          gamma Tubulin Antibody in Western Blot (WB)
          gamma Tubulin Antibody
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          gamma Tubulin Monoclonal Antibody (4D11)
          Product Details
          MA1-850
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          View 23 publications 23 publications
          Immunohistochemistry (IHC)
          -
          View 3 publications 3 publications
          Immunohistochemistry (Paraffin) (IHC (P))
          1:200
          -
          Immunocytochemistry (ICC/IF)
          1:50
          View 2 publications 2 publications
          Flow Cytometry (Flow)
          -
          View 1 publication 1 publication
          Immunoprecipitation (IP)
          Assay-dependent
          -
          Miscellaneous PubMed (Misc)
          -
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Human, Mouse, Non-human primate, Rat
          Published species
          Fruit fly, Human, Mouse, Non-human primate, Plant
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          4D11
          Immunogen
          Recombinant protein containing residues A A T R P D Y I S W G T Q E Q of human gamma Tubulin.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 555 
          • Alexa Fluor 647 
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS with 1mg/mL BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2211249
          Product Specific Information
          MA1-850 detects gamma Tubulin from human, mouse, monkey, and rat samples.
          MA1-850 has been successfully used in Western blot, immunoprecipitation, and immunofluorescence procedures.
          The MA1-850 immunogen is a recombinant protein containing residues A A T R P D Y I S W G T Q E Q of human gamma Tubulin.
          Target Information
          This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.
          Bioinformatics
          Protein Aliases: centrosome marker; gamma-1-tubulin; gamma-2-tubulin; gamma-tubulin complex component 1; GCP-1; TUBG1; tubulin gamma 2 chain; tubulin, gamma polypeptide; unnamed protein product
          View more View less
          Gene Aliases: 1500010O08Rik; CDCBM4; GCP-1; TUBG; TUBGCP1; Tubgl
          View more View less
          UniProt ID: (Human) P23258, (Human) Q9NRH3, (Rat) P83888, (Mouse) P83887, (Mouse) Q8VCK3
          View more View less
          Entrez Gene ID: (Human) 7283, (Human) 27175, (Rat) 252921, (Mouse) 103733, (Rat) 680991, (Mouse) 103768
          View more View less
          Function(s)
          structural constituent of cytoskeleton protein binding GTP binding identical protein binding microtubule nucleator activity structural molecule activity
          Process(es)
          mitotic sister chromatid segregation meiotic spindle organization mitotic cell cycle microtubule-based process microtubule nucleation mitotic spindle organization spermatid development cytoplasmic microtubule organization microtubule cytoskeleton organization regulation of mitotic nuclear division
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-blue-7fb9f846c9-wx6nt:80/100.66.72.150:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0