    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • Her-2 (ErbB2) Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Invitrogen
          ErbB2 (HER-2) Monoclonal Antibody (e2-4001), Biotin
          Advanced Verification
          3 Published Figures
          89 References
          View all (236) Her-2 (ErbB2) antibodies
          Tech Support
          Cite ErbB2 (HER-2) Monoclonal Antibody (e2-4001), Biotin
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          • Published Figures (3)
          • Advanced Verification (1)
          ErbB2 (HER-2) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ErbB2 (HER-2) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 8
          ErbB2 (HER-2) Antibody (MA5-13102) in WB
          Western blot analysis was performed on membrane enriched extracts (30 µg lysate) of SK-BR-3 (1) and MDA-MB-231 (2). The blots were probed with Anti-ErbB2 Mouse Monoclonal Antibody (Product # MA5-13102, 1 µg/mL dilution) and detected by chemiluminescence using Streptavidin-HRP (Product # N200, 1:10,000 dilution). A 185 kDa band corresponding to ErbB2 was observed in SK-BR-3 but not in MDA-MB-231, which is a negative control for ErbB2. Known quantity of protein samples were electrophoresed using Novex® NuPAGE® 4-12 % Bis-Tris gel (Product # ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ErbB2 (HER-2) Antibody in Western Blot (WB)
          ErbB2 (HER-2) Antibody in Western Blot (WB)
          ErbB2 (HER-2) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ErbB2 (HER-2) Antibody in Immunoprecipitation (IP)
          ErbB2 (HER-2) Antibody in Immunocytochemistry (ICC/IF)
          ErbB2 (HER-2) Antibody in Immunocytochemistry (ICC/IF)
          ErbB2 (HER-2) Antibody in Immunocytochemistry (ICC/IF)
          ErbB2 (HER-2) Antibody
          Product Details
          MA5-13102
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1-2 µg/mL
          View 24 publications 24 publications
          Immunohistochemistry (IHC)
          -
          View 51 publications 51 publications
          Immunohistochemistry (Paraffin) (IHC (P))
          1:200-1:400
          -
          Immunocytochemistry (ICC/IF)
          -
          View 6 publications 6 publications
          ELISA (ELISA)
          -
          View 1 publication 1 publication
          Immunoprecipitation (IP)
          2 µg/mL
          View 6 publications 6 publications
          Gel Shift (EMSA)
          -
          View 1 publication 1 publication
          Product Specifications
          Published species
          Cat, Hamster, Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          e2-4001
          Immunogen
          Cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein
          Conjugate
          Biotin Biotin Biotin
          Form
          Liquid
          Concentration
          0.2 mg/mL
          Amount
          100 µg
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_10984470
          Product Specific Information
          MA5-13102 targets HER-2 in IHC, IP, and WB applications and shows reactivity with Human, mouse, and Rat samples.
          The MA5-13102 immunogen is cytoplasmic domain of recombinant human c-erbB-2/HER-2 oncoprotein.
          Target Information
          ErbB2 (HER2) is a receptor tyrosine kinase that is overexpressed in some breast tumors. Herceptin, used in treatment of metastatic Her2-positive cancer, is monoclonal antibody targeting this kinase.
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline